missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(3-Bromothien-2-yl)-N-methylbenzylamine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC67846DE

 View more versions of this product

Catalog No. CC67846DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

4-(3-Bromothien-2-yl)-N-methylbenzylamine
96.5
97%
C12H12BrNS
5g
JAQWWPQIUYWZIC-UHFFFAOYSA-N
1-[4-(3-bromothiophen-2-yl)phenyl]-N-methylmethanamine
24229757
97%
937796-02-6
100.0
Amber Glass Bottle
MFCD09879974
4-3-bromothien-2-yl-n-methylbenzylamine, 4-3-bromothiophen-2-yl phenyl methyl methyl amine, 1-4-3-bromothiophen-2-yl phenyl-n-methylmethanamine, 4-3-bromothiophen-2-yl-n-methylbenzylamine
CNCC1=CC=C(C=C1)C2=C(C=CS2)Br
282.199
282.2

Chemical Identifiers

937796-02-6
282.199
JAQWWPQIUYWZIC-UHFFFAOYSA-N
24229757
CNCC1=CC=C(C=C1)C2=C(C=CS2)Br
C12H12BrNS
MFCD09879974
4-3-bromothien-2-yl-n-methylbenzylamine, 4-3-bromothiophen-2-yl phenyl methyl methyl amine, 1-4-3-bromothiophen-2-yl phenyl-n-methylmethanamine, 4-3-bromothiophen-2-yl-n-methylbenzylamine
1-[4-(3-bromothiophen-2-yl)phenyl]-N-methylmethanamine
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specifica

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions