missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Thienyl)benzoic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 29886-64-4
Molecular Formula C11H8O2S
Molecular Weight (g/mol) 204.243
MDL Number MFCD03783559
InChI Key FISAUHGRILVMDP-UHFFFAOYSA-N
Synonym 4-3-thienyl benzoic acid, 4-thiophen-3-yl benzoic acid, 4-thiophen-3-yl-benzoic acid, benzoic acid, 4-3-thienyl, 4-thien-3-ylbenzoic acid, 4-3-thienyl-benzoic acid, maybridge3_004545, acmc-1cisg
PubChem CID 736862
IUPAC Name 4-thiophen-3-ylbenzoic acid
SMILES C1=CC(=CC=C1C2=CSC=C2)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
MO07004CB
SDS View Documents
Thermo Scientific Chemicals
MO07004CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

29886-64-4
204.243
FISAUHGRILVMDP-UHFFFAOYSA-N
736862
C1=CC(=CC=C1C2=CSC=C2)C(=O)O
C11H8O2S
MFCD03783559
4-3-thienyl benzoic acid, 4-thiophen-3-yl benzoic acid, 4-thiophen-3-yl-benzoic acid, benzoic acid, 4-3-thienyl, 4-thien-3-ylbenzoic acid, 4-3-thienyl-benzoic acid, maybridge3_004545, acmc-1cisg
4-thiophen-3-ylbenzoic acid

Specifications

29886-64-4
97%
C11H8O2S
250mg
FISAUHGRILVMDP-UHFFFAOYSA-N
4-thiophen-3-ylbenzoic acid
736862
97%
97.0
Amber Glass Bottle
MFCD03783559
4-3-thienyl benzoic acid, 4-thiophen-3-yl benzoic acid, 4-thiophen-3-yl-benzoic acid, benzoic acid, 4-3-thienyl, 4-thien-3-ylbenzoic acid, 4-3-thienyl-benzoic acid, maybridge3_004545, acmc-1cisg
C1=CC(=CC=C1C2=CSC=C2)C(=O)O
204.243
204.25
4-(3-Thienyl)benzoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'