missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(3-Trifluoromethyl-2-pyridyloxy)benzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H33473MD

 View more versions of this product

Catalog No. AAH33473MD


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-(3-Trifluoromethyl-2-pyridyloxy)benzenesulfonyl chloride
63°C to 65°C
MFCD07774164
UN3261
FDAYBQVWHDJJOL-UHFFFAOYSA-N
4-[3-(trifluoromethyl)pyridin-2-yl]oxybenzenesulfonyl chloride
45594320
97%
874839-13-1
C12H7ClF3NO3S
250 mg
4-3-trifluoromethyl pyridin-2-yl oxy benzenesulfonyl chloride, 4-3-trifluoromethyl pyridin-2-yl oxy benzene-1-sulfonyl chloride, chloro 4-3-trifluoromethyl 2-pyridyloxy phenyl sulfone, 4-3-trifluoromethyl pyridin-2-yl oxy benzenesulphonyl chloride
C1=CC(=C(N=C1)OC2=CC=C(C=C2)S(=O)(=O)Cl)C(F)(F)F
337.697
337.71

Chemical Identifiers

874839-13-1
337.697
FDAYBQVWHDJJOL-UHFFFAOYSA-N
45594320
C1=CC(=C(N=C1)OC2=CC=C(C=C2)S(=O)(=O)Cl)C(F)(F)F
C12H7ClF3NO3S
MFCD07774164
4-3-trifluoromethyl pyridin-2-yl oxy benzenesulfonyl chloride, 4-3-trifluoromethyl pyridin-2-yl oxy benzene-1-sulfonyl chloride, chloro 4-3-trifluoromethyl 2-pyridyloxy phenyl sulfone, 4-3-trifluoromethyl pyridin-2-yl oxy benzenesulphonyl chloride
4-[3-(trifluoromethyl)pyridin-2-yl]oxybenzenesulfonyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only