missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzyl Bromide 95.0+%, TCI America™
Supplier: TCI America T28191G
Specifications
| 4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzyl Bromide | |
| C9H6BrF3N2 | |
| 1 g | |
| 3-[4-(Bromomethyl)phenyl]-3-(trifluoromethyl)-3H-diazirine | |
| FC(F)(F)C1(N=N1)C1=CC=C(CBr)C=C1 | |
| 279.06 | |
| 279.06 | |
| Liquid |
| 92367-11-8 | |
| MFCD19442581 | |
| 3265 | |
| BCFUQXOZHPIAPS-UHFFFAOYSA-N | |
| 3-[4-(bromomethyl)phenyl]-3-(trifluoromethyl)-3H-diazirine | |
| 10826496 | |
| ≥95.0% (HPLC) |
Chemical Identifiers
| 92367-11-8 | |
| 279.06 | |
| BCFUQXOZHPIAPS-UHFFFAOYSA-N | |
| 10826496 | |
| FC(F)(F)C1(N=N1)C1=CC=C(CBr)C=C1 |
| C9H6BrF3N2 | |
| MFCD19442581 | |
| 3-[4-(Bromomethyl)phenyl]-3-(trifluoromethyl)-3H-diazirine | |
| 3-[4-(bromomethyl)phenyl]-3-(trifluoromethyl)-3H-diazirine |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator