missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzylamine Hydrochloride 98.0+%, TCI America™
Supplier: TCI America T34481G
Specifications
4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzylamine Hydrochloride | |
142°C | |
C9H9ClF3N3 | |
3-[4-(Aminomethyl)phenyl]-3-(trifluoromethyl)-3H-diazirine Hydrochloride | |
C1=CC(=CC=C1CN)C2(N=N2)C(F)(F)F.Cl | |
251.637 | |
251.64 | |
Crystalline Powder |
1258874-29-1 | |
White | |
1 g | |
DNTDGTWJZOMTAK-UHFFFAOYSA-N | |
[4-[3-(trifluoromethyl)diazirin-3-yl]phenyl]methanamine;hydrochloride | |
66870186 | |
≥98.0% (HPLC,T) |
Chemical Identifiers
1258874-29-1 | |
251.637 | |
3-[4-(Aminomethyl)phenyl]-3-(trifluoromethyl)-3H-diazirine Hydrochloride | |
[4-[3-(trifluoromethyl)diazirin-3-yl]phenyl]methanamine;hydrochloride |
C9H9ClF3N3 | |
DNTDGTWJZOMTAK-UHFFFAOYSA-N | |
66870186 | |
C1=CC(=CC=C1CN)C2(N=N2)C(F)(F)F.Cl |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator