missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-{[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thien-2-yl]methyl}morpholine, 97%, Thermo Scientific™

$182.72 - $182.72

Chemical Identifiers

CAS 364794-85-4
Molecular Formula C15H24BNO3S
Molecular Weight (g/mol) 309.23
MDL Number MFCD11841079
InChI Key AMUMGAXTCHBNPU-UHFFFAOYSA-N
Synonym 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thien-2-ylmethyl morpholine, 4-4-tetramethyl-1,3,2-dioxaborolan-2-yl thiophen-2-yl methyl morpholine, 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophen-2-yl methyl morpholine, 2-morpholin-4-ylmethyl thiophene-4-boronic acid, pinacol ester, 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thien-2-yl methyl morpholine, amtb834, 2-morpholin-4-ylmethyl thiophene-4-boronic acid pinacol ester, 2-morpholin-4-ylmethyl thiophene-4-boronic acid,pinacol ester
PubChem CID 23438311
IUPAC Name 4-[[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]methyl]morpholine
SMILES CC1(C)OB(OC1(C)C)C1=CSC(CN2CCOCC2)=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC74239CB
SDS View Documents
Thermo Scientific Chemicals
CC74239CB
250 mg Amber Glass Bottle
Each for $182.72
 
Description

Description

Specifications

Chemical Identifiers

364794-85-4
309.23
AMUMGAXTCHBNPU-UHFFFAOYSA-N
23438311
CC1(C)OB(OC1(C)C)C1=CSC(CN2CCOCC2)=C1
C15H24BNO3S
MFCD11841079
4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thien-2-ylmethyl morpholine, 4-4-tetramethyl-1,3,2-dioxaborolan-2-yl thiophen-2-yl methyl morpholine, 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophen-2-yl methyl morpholine, 2-morpholin-4-ylmethyl thiophene-4-boronic acid, pinacol ester, 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thien-2-yl methyl morpholine, amtb834, 2-morpholin-4-ylmethyl thiophene-4-boronic acid pinacol ester, 2-morpholin-4-ylmethyl thiophene-4-boronic acid,pinacol ester
4-[[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]methyl]morpholine

Specifications

364794-85-4
100.0
Amber Glass Bottle
MFCD11841079
4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thien-2-ylmethyl morpholine, 4-4-tetramethyl-1,3,2-dioxaborolan-2-yl thiophen-2-yl methyl morpholine, 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophen-2-yl methyl morpholine, 2-morpholin-4-ylmethyl thiophene-4-boronic acid, pinacol ester, 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thien-2-yl methyl morpholine, amtb834, 2-morpholin-4-ylmethyl thiophene-4-boronic acid pinacol ester, 2-morpholin-4-ylmethyl thiophene-4-boronic acid,pinacol ester
CC1(C)OB(OC1(C)C)C1=CSC(CN2CCOCC2)=C1
309.23
309.24
4-{[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thien-2-yl]methyl}morpholine
96.5
97%
C15H24BNO3S
250 mg
AMUMGAXTCHBNPU-UHFFFAOYSA-N
4-[[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]methyl]morpholine
23438311
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'