missing translation for 'onlineSavingsMsg'
Learn More

4'-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)biphenyl-4-ol 98.0+%, TCI America™

Supplier:  TCI America T30311G

 View more versions of this product

Catalog No. T30311G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4′-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)biphenyl-4-ol
150°C
C18H21BO3
1 g
BFQSVDDJLRMXOO-UHFFFAOYSA-N
4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]phenol
69201437
≥98.0% (GC)
760989-91-1
White-Yellow
MFCD22414472
2-(4′C-Hydroxybiphenyl-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, (4′C-Hydroxybiphenyl-4-yl)boronic Acid Pinacol Ester
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C3=CC=C(C=C3)O
296.173
296.17
Crystalline Powder

Chemical Identifiers

760989-91-1
296.173
BFQSVDDJLRMXOO-UHFFFAOYSA-N
4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]phenol
C18H21BO3
MFCD22414472
69201437
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C3=CC=C(C=C3)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions