missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol 98.0+%, TCI America™
Supplier: TCI America T19541G
Specifications
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol | |
118°C | |
MFCD02093756 | |
4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 4-hydroxyphenylboronic acid pinacol ester, 4-hydroxyphenylboronic acid, pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 4-hydroxyphenylboronic acid pinacolate, 2-4-hydroxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-hydroxyphenylboronicacidpinacolester, 4-hydroxybenzeneboronic acid pinacol ester, phenol, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-hydroxyphenylboronic acid pinacol cyclic ester | |
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)O | |
220.075 | |
220.08 | |
Crystalline Powder |
269409-70-3 | |
C12H17BO3 | |
1 g | |
BICZJRAGTCRORZ-UHFFFAOYSA-N | |
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol | |
2734624 | |
≥98.0% (GC) |
Chemical Identifiers
269409-70-3 | |
220.075 | |
BICZJRAGTCRORZ-UHFFFAOYSA-N | |
2734624 | |
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)O |
C12H17BO3 | |
MFCD02093756 | |
4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 4-hydroxyphenylboronic acid pinacol ester, 4-hydroxyphenylboronic acid, pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 4-hydroxyphenylboronic acid pinacolate, 2-4-hydroxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-hydroxyphenylboronicacidpinacolester, 4-hydroxybenzeneboronic acid pinacol ester, phenol, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-hydroxyphenylboronic acid pinacol cyclic ester | |
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol |
Safety and Handling
TSCA : No