missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol 98.0+%, TCI America™

Supplier:  TCI America T19545G

 View more versions of this product

Catalog No. T19545G


Explore available promotions

Description

Description

Specifications

Specifications

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
118°C
MFCD02093756
4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 4-hydroxyphenylboronic acid pinacol ester, 4-hydroxyphenylboronic acid, pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 4-hydroxyphenylboronic acid pinacolate, 2-4-hydroxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-hydroxyphenylboronicacidpinacolester, 4-hydroxybenzeneboronic acid pinacol ester, phenol, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-hydroxyphenylboronic acid pinacol cyclic ester
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)O
220.075
220.08
Crystalline Powder
269409-70-3
C12H17BO3
5 g
BICZJRAGTCRORZ-UHFFFAOYSA-N
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
2734624
≥98.0% (GC)

Chemical Identifiers

269409-70-3
220.075
BICZJRAGTCRORZ-UHFFFAOYSA-N
2734624
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)O
C12H17BO3
MFCD02093756
4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 4-hydroxyphenylboronic acid pinacol ester, 4-hydroxyphenylboronic acid, pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 4-hydroxyphenylboronic acid pinacolate, 2-4-hydroxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-hydroxyphenylboronicacidpinacolester, 4-hydroxybenzeneboronic acid pinacol ester, phenol, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-hydroxyphenylboronic acid pinacol cyclic ester
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions