missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4,5,5,6,6,7,7,7-Nonafluoro-1-heptanol 98.0+%, TCI America™
$169.63 - $1737.30
Chemical Identifiers
| CAS | 83310-97-8 |
|---|---|
| Molecular Formula | C7H7F9O |
| Molecular Weight (g/mol) | 278.12 |
| MDL Number | MFCD02183552 |
| InChI Key | OVBNEUIFHDEQHD-UHFFFAOYSA-N |
| Synonym | 3-(Perfluorobutyl)propanol |
| PubChem CID | 2779091 |
| IUPAC Name | 4,4,5,5,6,6,7,7,7-nonafluoroheptan-1-ol |
| SMILES | OCCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)F |
Chemical Identifiers
| 83310-97-8 | |
| 278.12 | |
| OVBNEUIFHDEQHD-UHFFFAOYSA-N | |
| 2779091 | |
| OCCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)F |
| C7H7F9O | |
| MFCD02183552 | |
| 3-(Perfluorobutyl)propanol | |
| 4,4,5,5,6,6,7,7,7-nonafluoroheptan-1-ol |
Specifications
| 83310-97-8 | |
| 64°C | |
| MFCD02183552 | |
| 3-(Perfluorobutyl)propanol | |
| OCCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)F | |
| 278.12 | |
| 278.12 | |
| Liquid |
| Colorless | |
| C7H7F9O | |
| 5 g | |
| OVBNEUIFHDEQHD-UHFFFAOYSA-N | |
| 4,4,5,5,6,6,7,7,7-nonafluoroheptan-1-ol | |
| 2779091 | |
| ≥98.0% (GC) | |
| 4,4,5,5,6,6,7,7,7-Nonafluoro-1-heptanol |
Safety and Handling
TSCA : No