missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(4-Aminophenyl)thiomorpholine 1,1-Dioxide 98.0+%, TCI America™

$110.60 - $329.83

Chemical Identifiers

CAS 105297-10-7
Molecular Formula C10H14N2O2S
Molecular Weight (g/mol) 226.29
MDL Number MFCD06797046
InChI Key PLQZCPNIYKUNPR-UHFFFAOYSA-N
PubChem CID 26370384
IUPAC Name 4-(4-aminophenyl)-1λ⁶-thiomorpholine-1,1-dione
SMILES NC1=CC=C(C=C1)N1CCS(=O)(=O)CC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A16691G
View Documents
TCI America
A16691G
1 g
Each for $110.60
 
A16695G
View Documents
TCI America
A16695G
5 g
Each for $329.83
 
Description

Description

Specifications

Chemical Identifiers

105297-10-7
226.29
PLQZCPNIYKUNPR-UHFFFAOYSA-N
4-(4-aminophenyl)-1λ⁶-thiomorpholine-1,1-dione
C10H14N2O2S
MFCD06797046
26370384
NC1=CC=C(C=C1)N1CCS(=O)(=O)CC1

Specifications

105297-10-7
C10H14N2O2S
1 g
PLQZCPNIYKUNPR-UHFFFAOYSA-N
4-(4-aminophenyl)-1λ⁶-thiomorpholine-1,1-dione
26370384
≥98.0% (GC,T)
4-(4-Aminophenyl)thiomorpholine 1,1-Dioxide
194°C
MFCD06797046
4-(1,1-Dioxothiomorpholino)aniline, 4-Thiomorpholinoaniline 1′C,1′C-Dioxide
NC1=CC=C(C=C1)N1CCS(=O)(=O)CC1
226.29
226.29
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'