missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Azodianiline, 95%

CAS: 538-41-0 | C12H12N4 | 212.26 g/mol

$565.19 - $1939.73

Chemical Identifiers

CAS 538-41-0
Molecular Formula C12H12N4
Molecular Weight (g/mol) 212.26
MDL Number MFCD00041892
InChI Key KQIKKETXZQDHGE-UHFFFAOYSA-N
Synonym 4,4'-azodianiline, p-azoaniline, p-diaminoazobenzene, 4,4'-diaminoazobenzene, azodianiline, p'-amino-p-aminoazobenzene, 4,4'-azobisbenzenamine, benzenamine, 4,4'-azobis, 4,4-azodianiline, 4,4'-diazenediylbisaniline
PubChem CID 10855
IUPAC Name 4-[(4-aminophenyl)diazenyl]aniline
SMILES NC1=CC=C(C=C1)N=NC1=CC=C(N)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC401580050
SDS View Documents
Thermo Scientific Chemicals
401580050
5 g Glass bottle
Each for $565.19
Only null left
 
AC401580250
SDS View Documents
Thermo Scientific Chemicals
401580250
25 g Glass bottle
Each for $1,939.73
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

538-41-0
212.26
KQIKKETXZQDHGE-UHFFFAOYSA-N
10855
NC1=CC=C(C=C1)N=NC1=CC=C(N)C=C1
C12H12N4
MFCD00041892
4,4'-azodianiline, p-azoaniline, p-diaminoazobenzene, 4,4'-diaminoazobenzene, azodianiline, p'-amino-p-aminoazobenzene, 4,4'-azobisbenzenamine, benzenamine, 4,4'-azobis, 4,4-azodianiline, 4,4'-diazenediylbisaniline
4-[(4-aminophenyl)diazenyl]aniline

Specifications

538-41-0
100.0
Brown
95%
C12H12N4
5 g
4,4'-azodianiline, p-azoaniline, p-diaminoazobenzene, 4,4'-diaminoazobenzene, azodianiline, p'-amino-p-aminoazobenzene, 4,4'-azobisbenzenamine, benzenamine, 4,4'-azobis, 4,4-azodianiline, 4,4'-diazenediylbisaniline
KQIKKETXZQDHGE-UHFFFAOYSA-N
4-[(4-aminophenyl)diazenyl]aniline
10855
95%
4, 4'-Azodianiline, 95%
94.0
245°C
Authentic
Glass bottle
MFCD00041892
15, 2979
Solubility in water: slightly soluble. Other solubilities: soluble in ethanol,very soluble in benzene and chloroform
NC1=CC=C(C=C1)N=NC1=CC=C(N)C=C1
212.26
212.25
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Suspected of causing genetic defects.

GHS P Statement
IF exposed or concerned: Get medical advice/attention.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,ha

GHS Signal Word: Warning

EINECSNumber : 208-690-7

RTECSNumber : BW7450000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only