missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4,4'-Bis(3-aminophenoxy)diphenyl sulfone, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals A1719418
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4,4'-Bis(3-aminophenoxy)diphenyl sulfone | |
132°C to 135°C | |
MFCD00054738 | |
3,3'-sulfonylbis 4,1-phenylene bis oxy dianiline, bis 4-3-aminophenoxy phenyl sulfone, 4,4'-bis 3-aminophenoxy diphenyl sulfone, benzenamine, 3,3'-sulfonylbis 4,1-phenyleneoxy bis, 4,4-bis 3-aminophenoxy diphenyl sulfone, 3-4-4-3-aminophenoxy benzenesulfonyl phenoxy aniline, 4-3-aminophenoxy-1-4-3-aminophenoxy phenyl sulfonyl benzene, acmc-209hds, 3,3'-sulfonylbis 4,1-phenyleneoxy dianiline | |
NC1=CC(OC2=CC=C(C=C2)S(=O)(=O)C2=CC=C(OC3=CC(N)=CC=C3)C=C2)=CC=C1 | |
432.49 | |
432.5 |
30203-11-3 | |
C24H20N2O4S | |
50 g | |
WCXGOVYROJJXHA-UHFFFAOYSA-N | |
3-[4-[4-(3-aminophenoxy)phenyl]sulfonylphenoxy]aniline | |
121657 | |
95% |
Chemical Identifiers
30203-11-3 | |
432.49 | |
WCXGOVYROJJXHA-UHFFFAOYSA-N | |
121657 | |
NC1=CC(OC2=CC=C(C=C2)S(=O)(=O)C2=CC=C(OC3=CC(N)=CC=C3)C=C2)=CC=C1 |
C24H20N2O4S | |
MFCD00054738 | |
3,3'-sulfonylbis 4,1-phenylene bis oxy dianiline, bis 4-3-aminophenoxy phenyl sulfone, 4,4'-bis 3-aminophenoxy diphenyl sulfone, benzenamine, 3,3'-sulfonylbis 4,1-phenyleneoxy bis, 4,4-bis 3-aminophenoxy diphenyl sulfone, 3-4-4-3-aminophenoxy benzenesulfonyl phenoxy aniline, 4-3-aminophenoxy-1-4-3-aminophenoxy phenyl sulfonyl benzene, acmc-209hds, 3,3'-sulfonylbis 4,1-phenyleneoxy dianiline | |
3-[4-[4-(3-aminophenoxy)phenyl]sulfonylphenoxy]aniline |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 250-091-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only