missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4,4'-Bis(diethylphosphonomethyl)biphenyl 98.0+%, TCI America™
$48.79 - $68.98
Chemical Identifiers
CAS | 17919-34-5 |
---|---|
Molecular Formula | C22H32O6P2 |
Molecular Weight (g/mol) | 454.44 |
MDL Number | MFCD01321144 |
InChI Key | DRCOGQJUVZRNSQ-UHFFFAOYSA-N |
PubChem CID | 635132 |
IUPAC Name | 1-(diethoxyphosphorylmethyl)-4-[4-(diethoxyphosphorylmethyl)phenyl]benzene |
SMILES | CCOP(=O)(CC1=CC=C(C=C1)C2=CC=C(C=C2)CP(=O)(OCC)OCC)OCC |
Chemical Identifiers
17919-34-5 | |
454.44 | |
DRCOGQJUVZRNSQ-UHFFFAOYSA-N | |
1-(diethoxyphosphorylmethyl)-4-[4-(diethoxyphosphorylmethyl)phenyl]benzene |
C22H32O6P2 | |
MFCD01321144 | |
635132 | |
CCOP(=O)(CC1=CC=C(C=C1)C2=CC=C(C=C2)CP(=O)(OCC)OCC)OCC |
Specifications
17919-34-5 | |
C22H32O6P2 | |
1 g | |
DRCOGQJUVZRNSQ-UHFFFAOYSA-N | |
1-(diethoxyphosphorylmethyl)-4-[4-(diethoxyphosphorylmethyl)phenyl]benzene | |
635132 | |
≥98.0% (GC) | |
4,4′-Bis(diethylphosphonomethyl)biphenyl |
White | |
MFCD01321144 | |
[4,4′C-Biphenylylenebis(methylene)]bisphosphonic Acid Tetraethyl Ester, Tetraethyl [4,4′C-Biphenylylenebis(methylene)]bisphosphonate | |
CCOP(=O)(CC1=CC=C(C=C1)C2=CC=C(C=C2)CP(=O)(OCC)OCC)OCC | |
454.44 | |
454.44 | |
Crystalline Powder |
Safety and Handling
TSCA : No