missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(4-Bromo-1H-pyrazol-1-yl)benzenesulfonyl chloride, 95%, Thermo Scientific™

$222.19 - $222.19

Chemical Identifiers

CAS 1174064-62-0
Molecular Formula C9H6BrClN2O2S
Molecular Weight (g/mol) 321.573
MDL Number MFCD12406835
InChI Key LRYXAJMNOSSJDD-UHFFFAOYSA-N
Synonym 4-4-bromo-1h-pyrazol-1-yl benzenesulfonyl chloride, 4-4-bromopyrazol-1-yl benzenesulfonyl chloride, 4-4-bromo-1h-pyrazol-1-yl-benzenesulfonyl chloride, 4-4-bromo-1h-pyrazol-1-yl benzene-1-sulfonyl chloride
PubChem CID 45594324
IUPAC Name 4-(4-bromopyrazol-1-yl)benzenesulfonyl chloride
SMILES C1=CC(=CC=C1N2C=C(C=N2)Br)S(=O)(=O)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH33086MD
View Documents
Thermo Scientific Chemicals
H33086MD
250 mg
Each for $222.19
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1174064-62-0
321.573
LRYXAJMNOSSJDD-UHFFFAOYSA-N
45594324
C1=CC(=CC=C1N2C=C(C=N2)Br)S(=O)(=O)Cl
C9H6BrClN2O2S
MFCD12406835
4-4-bromo-1h-pyrazol-1-yl benzenesulfonyl chloride, 4-4-bromopyrazol-1-yl benzenesulfonyl chloride, 4-4-bromo-1h-pyrazol-1-yl-benzenesulfonyl chloride, 4-4-bromo-1h-pyrazol-1-yl benzene-1-sulfonyl chloride
4-(4-bromopyrazol-1-yl)benzenesulfonyl chloride

Specifications

1174064-62-0
C9H6BrClN2O2S
250 mg
Moisture sensitive
LRYXAJMNOSSJDD-UHFFFAOYSA-N
4-(4-bromopyrazol-1-yl)benzenesulfonyl chloride
45594324
95%
144°C to 146°C
MFCD12406835
UN3261
4-4-bromo-1h-pyrazol-1-yl benzenesulfonyl chloride, 4-4-bromopyrazol-1-yl benzenesulfonyl chloride, 4-4-bromo-1h-pyrazol-1-yl-benzenesulfonyl chloride, 4-4-bromo-1h-pyrazol-1-yl benzene-1-sulfonyl chloride
C1=CC(=CC=C1N2C=C(C=N2)Br)S(=O)(=O)Cl
321.573
321.58
4-(4-Bromo-1H-pyrazol-1-yl)benzenesulfonyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'