missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-(4-Bromo-3-fluorobenzyl)morpholine, 96%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H5998503
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-(4-Bromo-3-fluorobenzyl)morpholine | |
42°C to 43°C | |
MFCD15526848 | |
4-4-bromo-3-fluorobenzyl morpholine, n-4-bromo-3-fluorobenzyl morpholine, 4-4-bromo-3-fluorophenyl methyl morpholine, morpholine, 4-4-bromo-3-fluorophenyl methyl | |
FC1=C(Br)C=CC(CN2CCOCC2)=C1 | |
274.13 | |
274.13 |
897016-96-5 | |
C11H13BrFNO | |
1 g | |
AXLRZCPFGFYXIS-UHFFFAOYSA-N | |
4-[(4-bromo-3-fluorophenyl)methyl]morpholine | |
53417818 | |
96% |
Chemical Identifiers
897016-96-5 | |
274.13 | |
AXLRZCPFGFYXIS-UHFFFAOYSA-N | |
53417818 | |
FC1=C(Br)C=CC(CN2CCOCC2)=C1 |
C11H13BrFNO | |
MFCD15526848 | |
4-4-bromo-3-fluorobenzyl morpholine, n-4-bromo-3-fluorobenzyl morpholine, 4-4-bromo-3-fluorophenyl methyl morpholine, morpholine, 4-4-bromo-3-fluorophenyl methyl | |
4-[(4-bromo-3-fluorophenyl)methyl]morpholine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Keep cold; Store under Nitrogen