missing translation for 'onlineSavingsMsg'
Learn More

4'-(4-Bromophenyl)-2,2':6',2″-terpyridine 97.0+%, TCI America™

$186.41 - $659.87

Chemical Identifiers

CAS 89972-76-9
Molecular Formula C21H14BrN3
Molecular Weight (g/mol) 388.268
MDL Number MFCD06796984
InChI Key BOPPHKMZOYPASP-UHFFFAOYSA-N
Synonym 4'-4-bromophenyl-2,2':6',2-terpyridine, 4'-4-bromophenyl-2,6':2',2-terpyridine, 4'-4-bromophenyl-alpha,alpha',alpha-tripyridyl, 4-4-bromophenyl-6-pyridin-2-yl-2,2'-bipyridine, acmc-209r3y, 4'-4-bromophenyl-2,2':6',2'-terpyridine, 4-4'-bromophenyl-2,6-di 2-pyridyl pyridine, 4-4-bromophenyl-2,6-bis 2-pyridyl pyridine
PubChem CID 10762837
IUPAC Name 4-(4-bromophenyl)-2,6-dipyridin-2-ylpyridine
SMILES C1=CC=NC(=C1)C2=CC(=CC(=N2)C3=CC=CC=N3)C4=CC=C(C=C4)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B32181G
View Documents
TCI America
B32181G
1 g
Each for $186.41
Only null left
 
B32185G
View Documents
TCI America
B32185G
5 g
Each for $659.87
Only null left
 
Description

Description

Specifications

Chemical Identifiers

89972-76-9
388.268
BOPPHKMZOYPASP-UHFFFAOYSA-N
10762837
C1=CC=NC(=C1)C2=CC(=CC(=N2)C3=CC=CC=N3)C4=CC=C(C=C4)Br
C21H14BrN3
MFCD06796984
4'-4-bromophenyl-2,2':6',2-terpyridine, 4'-4-bromophenyl-2,6':2',2-terpyridine, 4'-4-bromophenyl-alpha,alpha',alpha-tripyridyl, 4-4-bromophenyl-6-pyridin-2-yl-2,2'-bipyridine, acmc-209r3y, 4'-4-bromophenyl-2,2':6',2'-terpyridine, 4-4'-bromophenyl-2,6-di 2-pyridyl pyridine, 4-4-bromophenyl-2,6-bis 2-pyridyl pyridine
4-(4-bromophenyl)-2,6-dipyridin-2-ylpyridine

Specifications

89972-76-9
White-Yellow
MFCD06796984
4'-4-bromophenyl-2,2':6',2-terpyridine, 4'-4-bromophenyl-2,6':2',2-terpyridine, 4'-4-bromophenyl-alpha,alpha',alpha-tripyridyl, 4-4-bromophenyl-6-pyridin-2-yl-2,2'-bipyridine, acmc-209r3y, 4'-4-bromophenyl-2,2':6',2'-terpyridine, 4-4'-bromophenyl-2,6-di 2-pyridyl pyridine, 4-4-bromophenyl-2,6-bis 2-pyridyl pyridine
C1=CC=NC(=C1)C2=CC(=CC(=N2)C3=CC=CC=N3)C4=CC=C(C=C4)Br
388.268
388.27
Crystalline Powder
162°C
C21H14BrN3
1 g
BOPPHKMZOYPASP-UHFFFAOYSA-N
4-(4-bromophenyl)-2,6-dipyridin-2-ylpyridine
10762837
≥97.0% (HPLC,T)
4′-(4-Bromophenyl)-2,2′:6′,2′′-terpyridine
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents