missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,4″-Diamino-p-terphenyl, 95%, Thermo Scientific Chemicals

$191.75 - $191.75

Chemical Identifiers

CAS 3365-85-3
Molecular Formula C18H16N2
Molecular Weight (g/mol) 260.34
MDL Number MFCD00051532
InChI Key QBSMHWVGUPQNJJ-UHFFFAOYSA-N
Synonym 4,4-diamino-p-terphenyl, 4,4-diaminoterphenyl, 4-4-4-aminophenyl phenyl aniline, ccris 4965, p-terphenyl, 4,4-diamine, 1,1':4',1-terphenyl-4,4-diamine, 4'-4-aminophenyl-1,1'-biphenyl-4-amine, 4,4-diamino-4-terphenyl
PubChem CID 104949
IUPAC Name 4-[4-(4-aminophenyl)phenyl]aniline
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)N)C3=CC=C(C=C3)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0540303
View Documents
Thermo Scientific Chemicals
L0540303
1 g
Each for $191.75
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3365-85-3
260.34
QBSMHWVGUPQNJJ-UHFFFAOYSA-N
104949
C1=CC(=CC=C1C2=CC=C(C=C2)N)C3=CC=C(C=C3)N
C18H16N2
MFCD00051532
4,4-diamino-p-terphenyl, 4,4-diaminoterphenyl, 4-4-4-aminophenyl phenyl aniline, ccris 4965, p-terphenyl, 4,4-diamine, 1,1':4',1-terphenyl-4,4-diamine, 4'-4-aminophenyl-1,1'-biphenyl-4-amine, 4,4-diamino-4-terphenyl
4-[4-(4-aminophenyl)phenyl]aniline

Specifications

3365-85-3
C18H16N2
1 g
4,4-diamino-p-terphenyl, 4,4-diaminoterphenyl, 4-4-4-aminophenyl phenyl aniline, ccris 4965, p-terphenyl, 4,4-diamine, 1,1':4',1-terphenyl-4,4-diamine, 4'-4-aminophenyl-1,1'-biphenyl-4-amine, 4,4-diamino-4-terphenyl
C1=CC(=CC=C1C2=CC=C(C=C2)N)C3=CC=C(C=C3)N
260.34
260.34
4,4''-Diamino-p-terphenyl
244°C to 250°C
MFCD00051532
2120295
QBSMHWVGUPQNJJ-UHFFFAOYSA-N
4-[4-(4-aminophenyl)phenyl]aniline
104949
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H341
Toxic if swallowed.
Suspected of causing genetic defects.

P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c

H302-H341

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only