missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Diaminoazobenzene, 95%

CAS: 538-41-0 | C12H12N4 | 212.26 g/mol

$115.27 - $967.38

Chemical Identifiers

CAS 538-41-0
Molecular Formula C12H12N4
Molecular Weight (g/mol) 212.26
MDL Number MFCD00041892
InChI Key KQIKKETXZQDHGE-UHFFFAOYSA-N
Synonym 4,4'-azodianiline, p-azoaniline, p-diaminoazobenzene, 4,4'-diaminoazobenzene, azodianiline, p'-amino-p-aminoazobenzene, 4,4'-azobisbenzenamine, benzenamine, 4,4'-azobis, 4,4-azodianiline, 4,4'-diazenediylbisaniline
PubChem CID 10855
SMILES NC1=CC=C(C=C1)N=NC1=CC=C(N)C=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL03743MD
View Documents
Thermo Scientific Chemicals
L03743MD
250 mg
Each for $115.27
Only null left
 
AAL0374303
View Documents
Thermo Scientific Chemicals
L0374303
1 g
Each for $285.99
Only null left
 
AAL0374306
View Documents
Thermo Scientific Chemicals
L0374306
5 g
Each for $967.38
Only null left
 
Description

Description

4,4'-Diaminoazobenzene is used in cross-linking studies of epoxy resins and the synthesis of surface relief holographic materials. It is used in synthesis of anthraquinone reactive dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Diaminoazobenzene is used in cross-linking studies of epoxy resins and the synthesis of surface relief holographic materials. It is used in synthesis of anthraquinone reactive dyes.

Solubility
Very soluble in ethanol. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

538-41-0
212.26
KQIKKETXZQDHGE-UHFFFAOYSA-N
10855
C12H12N4
MFCD00041892
4,4'-azodianiline, p-azoaniline, p-diaminoazobenzene, 4,4'-diaminoazobenzene, azodianiline, p'-amino-p-aminoazobenzene, 4,4'-azobisbenzenamine, benzenamine, 4,4'-azobis, 4,4-azodianiline, 4,4'-diazenediylbisaniline
NC1=CC=C(C=C1)N=NC1=CC=C(N)C=C1

Specifications

538-41-0
C12H12N4
250 mg
745553
4,4'-azodianiline, p-azoaniline, p-diaminoazobenzene, 4,4'-diaminoazobenzene, azodianiline, p'-amino-p-aminoazobenzene, 4,4'-azobisbenzenamine, benzenamine, 4,4'-azobis, 4,4-azodianiline, 4,4'-diazenediylbisaniline
KQIKKETXZQDHGE-UHFFFAOYSA-N
4-[(4-aminophenyl)diazenyl]aniline
10855
95%
∽245°C (decomposition)
MFCD00041892
UN2811
14,2977
Very soluble in ethanol. Insoluble in water.
NC1=CC=C(C=C1)N=NC1=CC=C(N)C=C1
212.26
212.26
4,4'-Diaminoazobenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c

H302+H312+H332-H341

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 208-690-7

RTECSNumber : BW7450000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only