missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,4'-Diaminostilbene-2,2'-disulfonic acid, 95%, Thermo Scientific Chemicals

$68.45 - $225.79

Chemical Identifiers

CAS 81-11-8
Molecular Formula C14H14N2O6S2
Molecular Weight (g/mol) 370.394
MDL Number MFCD00024946
InChI Key REJHVSOVQBJEBF-OWOJBTEDSA-N
Synonym amsonic acid, flavonic acid, tinopal bhs, amsonsaeure, dasd, 4,4'-diaminostilbene-2,2'-disulfonic acid, unii-h68088wvjl, 2,2'-disulfo-4,4'-stilbenediamine, diaminostilbenedisulfonic acid, 4,4'-diaminostilbene-2,2'-disulphonic acid
PubChem CID 5284378
IUPAC Name 5-amino-2-[(E)-2-(4-amino-2-sulfophenyl)ethenyl]benzenesulfonic acid
SMILES C1=CC(=C(C=C1N)S(=O)(=O)O)C=CC2=C(C=C(C=C2)N)S(=O)(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1666414
View Documents
Thermo Scientific Chemicals
A1666414
25 g
Each for $68.45
 
AAA1666422
View Documents
Thermo Scientific Chemicals
A1666422
100 g
Each for $225.79
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

81-11-8
370.394
REJHVSOVQBJEBF-OWOJBTEDSA-N
5284378
C1=CC(=C(C=C1N)S(=O)(=O)O)C=CC2=C(C=C(C=C2)N)S(=O)(=O)O
C14H14N2O6S2
MFCD00024946
amsonic acid, flavonic acid, tinopal bhs, amsonsaeure, dasd, 4,4'-diaminostilbene-2,2'-disulfonic acid, unii-h68088wvjl, 2,2'-disulfo-4,4'-stilbenediamine, diaminostilbenedisulfonic acid, 4,4'-diaminostilbene-2,2'-disulphonic acid
5-amino-2-[(E)-2-(4-amino-2-sulfophenyl)ethenyl]benzenesulfonic acid

Specifications

81-11-8
C14H14N2O6S2
25 g
14,595
REJHVSOVQBJEBF-OWOJBTEDSA-N
5-amino-2-[(E)-2-(4-amino-2-sulfophenyl)ethenyl]benzenesulfonic acid
5284378
95%
>300°C
MFCD00024946
629516
amsonic acid, flavonic acid, tinopal bhs, amsonsaeure, dasd, 4,4'-diaminostilbene-2,2'-disulfonic acid, unii-h68088wvjl, 2,2'-disulfo-4,4'-stilbenediamine, diaminostilbenedisulfonic acid, 4,4'-diaminostilbene-2,2'-disulphonic acid
C1=CC(=C(C=C1N)S(=O)(=O)O)C=CC2=C(C=C(C=C2)N)S(=O)(=O)O
370.394
370.41
4,4'-Diaminostilbene-2,2'-disulfonic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

EINECSNumber : 201-325-2

RTECSNumber : WJ6603000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only