Learn More
4,4'-Dibromobenzophenone, 98+%, Thermo Scientific Chemicals
$142.44 - $392.63
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAL0820003
|
Thermo Scientific Chemicals
L0820003 |
1 g |
Each for $142.44
|
|
||||||
AAL0820006
|
Thermo Scientific Chemicals
L0820006 |
5 g |
Each for $392.63
|
|
||||||
Description
4,4'-Dibromobenzophenone is used in the synthesis of high molecular weight poly(p-phenylene) derivatives through palladium-catalyzed polycondensation reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4,4′-Dibromobenzophenone is used in the synthesis of high molecular weight poly(p-phenylene) derivatives through palladium-catalyzed polycondensation reaction.
Solubility
Soluble in ethanol, 2-propanol, acetone and toluene. Miscible with water.
Notes
Light sensitive. Incompatible with strong oxidizing agents and strong acids.
Chemical Identifiers
3988-03-2 |
Specifications
3988-03-2 | |
C13H8Br2O | |
1 g | |
4,4'-dibromobenzophenone, bis 4-bromophenyl methanone, methanone, bis 4-bromophenyl, p,p'-dibromobenzophenone, bis p-bromophenyl ketone, 4,4-dibromobenzophenone, di4-bromophenyl ketone, bis-4-bromo-phenyl-methanone, zlchem 265, pubchem7494 | |
LFABNOYDEODDFX-UHFFFAOYSA-N | |
bis(4-bromophenyl)methanone | |
72868 | |
≥98% |
172°C to 176°C | |
MFCD00016330 | |
2049958 | |
Soluble in ethanol,2-propanol,acetone and toluene. Miscible with water. | |
BrC1=CC=C(C=C1)C(=O)C1=CC=C(Br)C=C1 | |
340.01 | |
340.03 | |
4,4'-Dibromobenzophenone |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 223-632-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only