missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dibromobiphenyl, 99%

CAS: 92-86-4 | C12H8Br2 | 312.00 g/mol

$68.52 - $558.06

Chemical Identifiers

CAS 92-86-4
Molecular Formula C12H8Br2
Molecular Weight (g/mol) 312.00
MDL Number MFCD00000101
InChI Key HQJQYILBCQPYBI-UHFFFAOYSA-N
Synonym 4,4'-dibromobiphenyl, 1,1'-biphenyl, 4,4'-dibromo, p,p'-dibromobiphenyl, 4,4'-dibromodiphenyl, 1-bromo-4-4-bromophenyl benzene, 4,4'-dibromo-biphenyl, unii-pou26v2xt2, 4',4-dibromobiphenyl, biphenyl, 4,4'-dibromo, pou26v2xt2
PubChem CID 7110
IUPAC Name 1-bromo-4-(4-bromophenyl)benzene
SMILES BrC1=CC=C(C=C1)C1=CC=C(Br)C=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC212080250
SDS View Documents
Thermo Scientific Chemicals
212080250
25 g Plastic bottle
Each for $68.52
Only null left
 
AC212081000
SDS View Documents
Thermo Scientific Chemicals
212081000
100 g Plastic bottle
Each for $153.92
Only null left
 
AC212085000
SDS View Documents
Thermo Scientific Chemicals
212085000
500 g Plastic bottle
Each for $558.06
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

92-86-4
312.00
HQJQYILBCQPYBI-UHFFFAOYSA-N
7110
BrC1=CC=C(C=C1)C1=CC=C(Br)C=C1
C12H8Br2
MFCD00000101
4,4'-dibromobiphenyl, 1,1'-biphenyl, 4,4'-dibromo, p,p'-dibromobiphenyl, 4,4'-dibromodiphenyl, 1-bromo-4-4-bromophenyl benzene, 4,4'-dibromo-biphenyl, unii-pou26v2xt2, 4',4-dibromobiphenyl, biphenyl, 4,4'-dibromo, pou26v2xt2
1-bromo-4-(4-bromophenyl)benzene

Specifications

92-86-4
100.0
355°C to 360°C
Authentic
Plastic bottle
BrC6H4C6H4Br
25 g
4,4'-dibromobiphenyl, 1,1'-biphenyl, 4,4'-dibromo, p,p'-dibromobiphenyl, 4,4'-dibromodiphenyl, 1-bromo-4-4-bromophenyl benzene, 4,4'-dibromo-biphenyl, unii-pou26v2xt2, 4',4-dibromobiphenyl, biphenyl, 4,4'-dibromo, pou26v2xt2
HQJQYILBCQPYBI-UHFFFAOYSA-N
1-bromo-4-(4-bromophenyl)benzene
7110
99%
4, 4'-Dibromobiphenyl, 99%
98.5
165°C to 169°C
>100°C
98.5% min. (GC)
C12H8Br2
MFCD00000101
05, 580
Solubility in water: insoluble. Other solubilities: slightly soluble in ethanol;,soluble in benzene
BrC1=CC=C(C=C1)C1=CC=C(Br)C=C1
312.00
312
Crystalline Powder or Crystals
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-198-6

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only