missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4,4'-Dichlorochalcone, 98+%, Thermo Scientific Chemicals
$86.96 - $277.19
Chemical Identifiers
CAS | 19672-59-4 |
---|---|
Molecular Formula | C15H10Cl2O |
Molecular Weight (g/mol) | 277.14 |
MDL Number | MFCD00018704 |
InChI Key | YMEMCRBNZSLQCQ-XCVCLJGOSA-N |
Synonym | 4,4'-dichlorochalcone, e-1,3-bis-4-chloro-phenyl-propenone, e-1,3-bis 4-chlorophenyl prop-2-en-1-one, 2e-1,3-bis 4-chlorophenyl prop-2-en-1-one, 2e-1,3-bis 4-chlorophenyl-2-propen-1-one, 4',4-dichlorochalcone, e-4,4'-dichlorochalcone, trans-1,3-di-4-chlorophenyl-prop-2-en-1-one |
PubChem CID | 5377011 |
IUPAC Name | (E)-1,3-bis(4-chlorophenyl)prop-2-en-1-one |
SMILES | ClC1=CC=C(\C=C\C(=O)C2=CC=C(Cl)C=C2)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1071706
|
Thermo Scientific Chemicals
L1071706 |
5 g |
Each for $86.96
|
|
|||||
AAL1071714
|
Thermo Scientific Chemicals
L1071714 |
25 g |
Each for $277.19
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
19672-59-4 | |
277.14 | |
YMEMCRBNZSLQCQ-XCVCLJGOSA-N | |
5377011 | |
ClC1=CC=C(\C=C\C(=O)C2=CC=C(Cl)C=C2)C=C1 |
C15H10Cl2O | |
MFCD00018704 | |
4,4'-dichlorochalcone, e-1,3-bis-4-chloro-phenyl-propenone, e-1,3-bis 4-chlorophenyl prop-2-en-1-one, 2e-1,3-bis 4-chlorophenyl prop-2-en-1-one, 2e-1,3-bis 4-chlorophenyl-2-propen-1-one, 4',4-dichlorochalcone, e-4,4'-dichlorochalcone, trans-1,3-di-4-chlorophenyl-prop-2-en-1-one | |
(E)-1,3-bis(4-chlorophenyl)prop-2-en-1-one |
Specifications
19672-59-4 | |
C15H10Cl2O | |
5 g | |
4,4'-dichlorochalcone, e-1,3-bis-4-chloro-phenyl-propenone, e-1,3-bis 4-chlorophenyl prop-2-en-1-one, 2e-1,3-bis 4-chlorophenyl prop-2-en-1-one, 2e-1,3-bis 4-chlorophenyl-2-propen-1-one, 4',4-dichlorochalcone, e-4,4'-dichlorochalcone, trans-1,3-di-4-chlorophenyl-prop-2-en-1-one | |
ClC1=CC=C(\C=C\C(=O)C2=CC=C(Cl)C=C2)C=C1 | |
277.14 | |
277.15 | |
4,4'-Dichlorochalcone |
156°C to 160°C | |
MFCD00018704 | |
2331589 | |
YMEMCRBNZSLQCQ-XCVCLJGOSA-N | |
(E)-1,3-bis(4-chlorophenyl)prop-2-en-1-one | |
5377011 | |
≥98% |
Safety and Handling
RTECSNumber : UD5572635
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only