missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Diethynylbiphenyl 98.0+%, TCI America™
$227.41 - $794.10
Chemical Identifiers
| CAS | 38215-38-2 |
|---|---|
| Molecular Formula | C16H10 |
| Molecular Weight (g/mol) | 202.26 |
| MDL Number | MFCD00458964 |
| InChI Key | MXJJMQSKDPNPSX-UHFFFAOYSA-N |
| PubChem CID | 10013083 |
| IUPAC Name | 4,4'-diethynyl-1,1'-biphenyl |
| SMILES | C#CC1=CC=C(C=C1)C1=CC=C(C=C1)C#C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D4233200MG
|
TCI America
D4233200MG |
200 mg |
Each for $227.41
|
|
|||||
|
D42331G
|
TCI America
D42331G |
1 g |
Each for $794.10
|
|
|||||
Chemical Identifiers
| 38215-38-2 | |
| 202.26 | |
| MXJJMQSKDPNPSX-UHFFFAOYSA-N | |
| 4,4'-diethynyl-1,1'-biphenyl |
| C16H10 | |
| MFCD00458964 | |
| 10013083 | |
| C#CC1=CC=C(C=C1)C1=CC=C(C=C1)C#C |
Specifications
| 38215-38-2 | |
| Yellow | |
| MFCD00458964 | |
| MXJJMQSKDPNPSX-UHFFFAOYSA-N | |
| 4,4'-diethynyl-1,1'-biphenyl | |
| 10013083 | |
| ≥98.0% (GC) | |
| 4,4′-Diethynylbiphenyl |
| 168°C | |
| C16H10 | |
| 200 mg | |
| C#CC1=CC=C(C=C1)C1=CC=C(C=C1)C#C | |
| 202.26 | |
| 202.26 | |
| Crystalline Powder |
Safety and Handling
TSCA : No