missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(4,4-Difluorocyclohexyl)methanol, 97%, Thermo Scientific™

Chemical Identifiers

CAS 178312-48-6
Molecular Formula C7H12F2O
Molecular Weight (g/mol) 150.17
MDL Number MFCD11847775
InChI Key XJZNZSLOHZLFQP-UHFFFAOYSA-N
Synonym 4,4-difluorocyclohexyl methanol, 4,4-difluoro-cyclohexyl-methanol, 4,4-difluorocyclohexane-1-methanol, cyclohexanemethanol, 4,4-difluoro, 4,4-difluorocyclohexanemethanol, 4,4-difluoro cyclohexyl methanol, 4,4-difluoro-cyclohexyl methanol, 4,4-difluorocyclohexyl-methanol
PubChem CID 21184133
IUPAC Name (4,4-difluorocyclohexyl)methanol
SMILES OCC1CCC(F)(F)CC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC452470010
SDS View Documents
Thermo Scientific Chemicals
452470010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC452470050
SDS View Documents
Thermo Scientific Chemicals
452470050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

178312-48-6
150.17
XJZNZSLOHZLFQP-UHFFFAOYSA-N
21184133
OCC1CCC(F)(F)CC1
C7H12F2O
MFCD11847775
4,4-difluorocyclohexyl methanol, 4,4-difluoro-cyclohexyl-methanol, 4,4-difluorocyclohexane-1-methanol, cyclohexanemethanol, 4,4-difluoro, 4,4-difluorocyclohexanemethanol, 4,4-difluoro cyclohexyl methanol, 4,4-difluoro-cyclohexyl methanol, 4,4-difluorocyclohexyl-methanol
(4,4-difluorocyclohexyl)methanol

Specifications

178312-48-6
C7H12F2O
1g
XJZNZSLOHZLFQP-UHFFFAOYSA-N
(4,4-difluorocyclohexyl)methanol
21184133
97%
96% min. (GC)
MFCD11847775
4,4-difluorocyclohexyl methanol, 4,4-difluoro-cyclohexyl-methanol, 4,4-difluorocyclohexane-1-methanol, cyclohexanemethanol, 4,4-difluoro, 4,4-difluorocyclohexanemethanol, 4,4-difluoro cyclohexyl methanol, 4,4-difluoro-cyclohexyl methanol, 4,4-difluorocyclohexyl-methanol
OCC1CCC(F)(F)CC1
150.17
150.17
(4, 4-Difluorocyclohexyl)methanol
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only