missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4,4'-Dihydroxy-3,3',5,5'-tetraisopropylbiphenyl 97.0+%, TCI America™
Supplier: TCI America D52811G
Specifications
4,4′-Dihydroxy-3,3′,5,5′-tetraisopropylbiphenyl | |
109°C | |
MFCD20257818 | |
3,3′,5,5′-Tetraisopropyl-[1,1′-biphenyl]-4,4′-diol | |
CC(C)C1=CC(=CC(C(C)C)=C1O)C1=CC(C(C)C)=C(O)C(=C1)C(C)C | |
354.53 | |
354.53 | |
Crystalline Powder |
2416-95-7 | |
C24H34O2 | |
1 g | |
QAISRHCMPQROAX-UHFFFAOYSA-N | |
3,3',5,5'-tetrakis(propan-2-yl)-[1,1'-biphenyl]-4,4'-diol | |
11602828 | |
≥97.0% (GC) |
Chemical Identifiers
2416-95-7 | |
354.53 | |
QAISRHCMPQROAX-UHFFFAOYSA-N | |
11602828 | |
CC(C)C1=CC(=CC(C(C)C)=C1O)C1=CC(C(C)C)=C(O)C(=C1)C(C)C |
C24H34O2 | |
MFCD20257818 | |
3,3′,5,5′-Tetraisopropyl-[1,1′-biphenyl]-4,4′-diol | |
3,3',5,5'-tetrakis(propan-2-yl)-[1,1'-biphenyl]-4,4'-diol |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator