missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Dihydroxytetraphenylmethane 98.0+%, TCI America™
$85.75 - $369.08
Chemical Identifiers
| CAS | 1844-01-5 |
|---|---|
| Molecular Formula | C25H20O2 |
| Molecular Weight (g/mol) | 352.433 |
| MDL Number | MFCD00045785 |
| InChI Key | BATCUENAARTUKW-UHFFFAOYSA-N |
| Synonym | Bis(4-hydroxyphenyl)diphenylmethane, (Diphenyl)bis(4-hydroxyphenyl)methane |
| PubChem CID | 232881 |
| IUPAC Name | 4-[(4-hydroxyphenyl)-diphenylmethyl]phenol |
| SMILES | C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)O |
Chemical Identifiers
| 1844-01-5 | |
| 352.433 | |
| BATCUENAARTUKW-UHFFFAOYSA-N | |
| 232881 | |
| C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)O |
| C25H20O2 | |
| MFCD00045785 | |
| Bis(4-hydroxyphenyl)diphenylmethane, (Diphenyl)bis(4-hydroxyphenyl)methane | |
| 4-[(4-hydroxyphenyl)-diphenylmethyl]phenol |
Specifications
| 1844-01-5 | |
| C25H20O2 | |
| 5 g | |
| BATCUENAARTUKW-UHFFFAOYSA-N | |
| 4-[(4-hydroxyphenyl)-diphenylmethyl]phenol | |
| 232881 | |
| ≥98.0% (GC) | |
| 4,4′-Dihydroxytetraphenylmethane |
| White-Yellow | |
| MFCD00045785 | |
| Bis(4-hydroxyphenyl)diphenylmethane, (Diphenyl)bis(4-hydroxyphenyl)methane | |
| C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)O | |
| 352.433 | |
| 352.43 | |
| Crystalline Powder |
Safety and Handling
TSCA : No