missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4,4-Dimethyl-2-cyclohexen-1-one 96.0+%, TCI America™
$248.86 - $989.48
Chemical Identifiers
CAS | 1073-13-8 |
---|---|
Molecular Formula | C8H12O |
Molecular Weight (g/mol) | 124.18 |
MDL Number | MFCD00009695 |
InChI Key | HAUNPYVLVAIUOO-UHFFFAOYSA-N |
Synonym | 4,4-dimethyl-2-cyclohexen-1-one, 4,4-dimethylcyclohex-2-enone, 2-cyclohexen-1-one, 4,4-dimethyl, 4,4-dimethyl-2-cyclohexenone, 4,4-dimethylcyclohexenone, 4,4-dimethyl-2-cyclohexene-1-one, 4,4-dimethyl-cyclohex-2-en-1-one, pubchem17152, acmc-1bxzo, 3,3-dimethylcyclohexene-6-one |
PubChem CID | 136839 |
IUPAC Name | 4,4-dimethylcyclohex-2-en-1-one |
SMILES | CC1(C)CCC(=O)C=C1 |
Chemical Identifiers
1073-13-8 | |
124.18 | |
HAUNPYVLVAIUOO-UHFFFAOYSA-N | |
136839 | |
CC1(C)CCC(=O)C=C1 |
C8H12O | |
MFCD00009695 | |
4,4-dimethyl-2-cyclohexen-1-one, 4,4-dimethylcyclohex-2-enone, 2-cyclohexen-1-one, 4,4-dimethyl, 4,4-dimethyl-2-cyclohexenone, 4,4-dimethylcyclohexenone, 4,4-dimethyl-2-cyclohexene-1-one, 4,4-dimethyl-cyclohex-2-en-1-one, pubchem17152, acmc-1bxzo, 3,3-dimethylcyclohexene-6-one | |
4,4-dimethylcyclohex-2-en-1-one |
Specifications
1073-13-8 | |
74°C | |
MFCD00009695 | |
4,4-dimethyl-2-cyclohexen-1-one, 4,4-dimethylcyclohex-2-enone, 2-cyclohexen-1-one, 4,4-dimethyl, 4,4-dimethyl-2-cyclohexenone, 4,4-dimethylcyclohexenone, 4,4-dimethyl-2-cyclohexene-1-one, 4,4-dimethyl-cyclohex-2-en-1-one, pubchem17152, acmc-1bxzo, 3,3-dimethylcyclohexene-6-one | |
CC1(C)CCC(=O)C=C1 | |
124.18 | |
124.18 | |
Liquid |
Yellow | |
C8H12O | |
5 g | |
HAUNPYVLVAIUOO-UHFFFAOYSA-N | |
4,4-dimethylcyclohex-2-en-1-one | |
136839 | |
≥96.0% (GC) | |
4,4-Dimethyl-2-cyclohexen-1-one |
Safety and Handling
TSCA : No