missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Dimethyl-trans-stilbene 99.0+%, TCI America™
Supplier: TCI America D27085G
Specifications
| 4,4′-Dimethyl-trans-stilbene | |
| 183°C | |
| C16H16 | |
| 5 g | |
| CC1=CC=C(C=C1)C=CC2=CC=C(C=C2)C | |
| 208.304 | |
| 208.30 | |
| Crystalline Powder |
| 18869-29-9 | |
| White | |
| MFCD00136904 | |
| KINZBJFIDFZQCB-VAWYXSNFSA-N | |
| 1-methyl-4-[(E)-2-(4-methylphenyl)ethenyl]benzene | |
| 5462964 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 18869-29-9 | |
| 208.304 | |
| KINZBJFIDFZQCB-VAWYXSNFSA-N | |
| 1-methyl-4-[(E)-2-(4-methylphenyl)ethenyl]benzene |
| C16H16 | |
| MFCD00136904 | |
| 5462964 | |
| CC1=CC=C(C=C1)C=CC2=CC=C(C=C2)C |
Safety and Handling
TSCA : No