missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4,4'-Dinitrostilbene 98.0+%, TCI America™
$537.21 - $1923.65
Chemical Identifiers
CAS | 2501-02-2 |
---|---|
Molecular Formula | C14H10N2O4 |
Molecular Weight (g/mol) | 270.24 |
MDL Number | MFCD00017043 |
InChI Key | CLVIIRIMEIEKOQ-OWOJBTEDSA-N |
PubChem CID | 5377860 |
ChEBI | CHEBI:79798 |
IUPAC Name | 1-nitro-4-[(E)-2-(4-nitrophenyl)ethenyl]benzene |
SMILES | C1=CC(=CC=C1C=CC2=CC=C(C=C2)[N+](=O)[O-])[N+](=O)[O-] |
Chemical Identifiers
2501-02-2 | |
270.24 | |
CLVIIRIMEIEKOQ-OWOJBTEDSA-N | |
CHEBI:79798 | |
C1=CC(=CC=C1C=CC2=CC=C(C=C2)[N+](=O)[O-])[N+](=O)[O-] |
C14H10N2O4 | |
MFCD00017043 | |
5377860 | |
1-nitro-4-[(E)-2-(4-nitrophenyl)ethenyl]benzene |
Specifications
2501-02-2 | |
Yellow | |
MFCD00017043 | |
CLVIIRIMEIEKOQ-OWOJBTEDSA-N | |
1-nitro-4-[(E)-2-(4-nitrophenyl)ethenyl]benzene | |
5377860 | |
270.24 | |
Crystalline Powder |
295°C | |
C14H10N2O4 | |
5 g | |
C1=CC(=CC=C1C=CC2=CC=C(C=C2)[N+](=O)[O-])[N+](=O)[O-] | |
270.24 | |
CHEBI:79798 | |
≥98.0% (HPLC) | |
4,4′-Dinitrostilbene |
Safety and Handling
TSCA : No