missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4,4'-(Ethyne-1,2-diyl)diphthalic Anhydride 98.0+%, TCI America™
Supplier: TCI America E11645G
Specifications
4,4′-(Ethyne-1,2-diyl)diphthalic Anhydride | |
350°C | |
5 g | |
XGZRRDYHYZLYIJ-UHFFFAOYSA-N | |
5-[2-(1,3-dioxo-2-benzofuran-5-yl)ethynyl]-2-benzofuran-1,3-dione | |
58986478 | |
≥98.0% (HPLC,T) |
129808-00-0 | |
C18H6O6 | |
5,5′C-(Ethyne-1,2-diyl)bis(isobenzofuran-1,3-dione) | |
C1=CC2=C(C=C1C#CC3=CC4=C(C=C3)C(=O)OC4=O)C(=O)OC2=O | |
318.24 | |
318.24 | |
Crystalline Powder |
Chemical Identifiers
129808-00-0 | |
318.24 | |
58986478 | |
C1=CC2=C(C=C1C#CC3=CC4=C(C=C3)C(=O)OC4=O)C(=O)OC2=O |
C18H6O6 | |
XGZRRDYHYZLYIJ-UHFFFAOYSA-N | |
5-[2-(1,3-dioxo-2-benzofuran-5-yl)ethynyl]-2-benzofuran-1,3-dione |
Safety and Handling
TSCA : No