missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(4-Fluorobenzoyl)pyridine, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2232706

 View more versions of this product

Catalog No. AAB2232706


Explore available promotions

Description

Description

4-(4-Fluorobenzoyl)pyridine is used for increasing cardiac contractility and for the treatment of congestive heart failure, and pharmaceutical compositions and for the preparation thereof.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(4-Fluorobenzoyl)pyridine is used for increasing cardiac contractility and for the treatment of congestive heart failure, and pharmaceutical compositions and for the preparation thereof.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

4-(4-Fluorobenzoyl)pyridine
85°C to 87°C
MFCD02683088
4-4-fluorobenzoyl pyridine, 4-fluorophenyl pyridin-4-yl methanone, methanone, 4-fluorophenyl-4-pyridinyl, 4-fluorophenyl-pyridin-4-ylmethanone, 4-fluorophenyl 4-pyridyl ketone, acmc-20amw5, d0j0wo, 4-pyridyl p-fluorophenyl ketone, 4-4-fluorophenyl carbonyl pyridine, 4-fluorophenyl 4-pyridinyl methanone
FC1=CC=C(C=C1)C(=O)C1=CC=NC=C1
201.20
201.2
41538-36-7
C12H8FNO
5 g
WTRWBYGUMQEFFI-UHFFFAOYSA-N
(4-fluorophenyl)-pyridin-4-ylmethanone
7023019
99%

Chemical Identifiers

41538-36-7
201.20
WTRWBYGUMQEFFI-UHFFFAOYSA-N
7023019
C12H8FNO
MFCD02683088
4-4-fluorobenzoyl pyridine, 4-fluorophenyl pyridin-4-yl methanone, methanone, 4-fluorophenyl-4-pyridinyl, 4-fluorophenyl-pyridin-4-ylmethanone, 4-fluorophenyl 4-pyridyl ketone, acmc-20amw5, d0j0wo, 4-pyridyl p-fluorophenyl ketone, 4-4-fluorophenyl carbonyl pyridine, 4-fluorophenyl 4-pyridinyl methanone
FC1=CC=C(C=C1)C(=O)C1=CC=NC=C1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only