missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(4-Fluorophenoxy)benzaldehyde, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H50452MD

 View more versions of this product

Catalog No. AAH50452MD



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-(4-Fluorophenoxy)benzaldehyde
C13H9FO2
250 mg
YUPBWHURNLRZQL-UHFFFAOYSA-N
4-(4-fluorophenoxy)benzaldehyde
3856802
97%
137736-06-2
MFCD01631896
4-4-fluorophenoxy benzaldehyde, 4-4'-fluorophenoxy benzaldehyde, 4-4-fluoro-phenoxy-benzaldehyde, benzaldehyde, 4-4-fluorophenoxy, pubchem23015, acmc-209ceq, yupbwhurnlrzql-uhfffaoysa, 4-4-fluorophenoxyl benzaldehyde, 4-fluoro-4'-formyldiphenyl ether
FC1=CC=C(OC2=CC=C(C=O)C=C2)C=C1
216.21
216.21

Chemical Identifiers

137736-06-2
216.21
YUPBWHURNLRZQL-UHFFFAOYSA-N
3856802
FC1=CC=C(OC2=CC=C(C=O)C=C2)C=C1
C13H9FO2
MFCD01631896
4-4-fluorophenoxy benzaldehyde, 4-4'-fluorophenoxy benzaldehyde, 4-4-fluoro-phenoxy-benzaldehyde, benzaldehyde, 4-4-fluorophenoxy, pubchem23015, acmc-209ceq, yupbwhurnlrzql-uhfffaoysa, 4-4-fluorophenoxyl benzaldehyde, 4-fluoro-4'-formyldiphenyl ether
4-(4-fluorophenoxy)benzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only