missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(4-Isocyanatopyrid-2-yl)morpholine, ≥90%, Thermo Scientific™

Chemical Identifiers

CAS 876316-43-7
Molecular Formula C10H11N3O2
Molecular Weight (g/mol) 205.22
MDL Number MFCD08271914
InChI Key LRVCWTODNVVIOK-UHFFFAOYSA-N
Synonym 4-4-isocyanatopyrid-2-yl morpholine, 4-4-isocyanatopyridin-2-yl morpholine, morpholine,4-4-isocyanato-2-pyridinyl, 2-morpholin-4-ylpyridin-4-isocyanate, 4-4-isocyanatopyrid-2-yl-morpholine, 2-morpholin-4-yl pyridin-4-yl isocyanate, 4-isocyanato-2-morpholin-4-yl pyridine
PubChem CID 18525762
IUPAC Name 4-(4-isocyanatopyridin-2-yl)morpholine
SMILES O=C=NC1=CC(=NC=C1)N1CCOCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC43106CB
SDS View Documents
Thermo Scientific Chemicals
CC43106CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC43106DA
SDS View Documents
Thermo Scientific Chemicals
CC43106DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

876316-43-7
205.22
LRVCWTODNVVIOK-UHFFFAOYSA-N
18525762
O=C=NC1=CC(=NC=C1)N1CCOCC1
C10H11N3O2
MFCD08271914
4-4-isocyanatopyrid-2-yl morpholine, 4-4-isocyanatopyridin-2-yl morpholine, morpholine,4-4-isocyanato-2-pyridinyl, 2-morpholin-4-ylpyridin-4-isocyanate, 4-4-isocyanatopyrid-2-yl-morpholine, 2-morpholin-4-yl pyridin-4-yl isocyanate, 4-isocyanato-2-morpholin-4-yl pyridine
4-(4-isocyanatopyridin-2-yl)morpholine

Specifications

876316-43-7
100.0
Amber Glass Bottle
MFCD08271914
4-4-isocyanatopyrid-2-yl morpholine, 4-4-isocyanatopyridin-2-yl morpholine, morpholine,4-4-isocyanato-2-pyridinyl, 2-morpholin-4-ylpyridin-4-isocyanate, 4-4-isocyanatopyrid-2-yl-morpholine, 2-morpholin-4-yl pyridin-4-yl isocyanate, 4-isocyanato-2-morpholin-4-yl pyridine
O=C=NC1=CC(=NC=C1)N1CCOCC1
205.22
205.22
4-(4-Isocyanatopyrid-2-yl)morpholine
94.5
95%
C10H11N3O2
250mg
LRVCWTODNVVIOK-UHFFFAOYSA-N
4-(4-isocyanatopyridin-2-yl)morpholine
18525762
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'