missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(4-Isopropylpiperazin-1-yl)phenylboronic acid pinacol ester, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 445080010

 View more versions of this product

Catalog No. AC445080010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

4-(4-Isopropylpiperazin-1-yl)phenylboronic acid pinacol ester
96% min. (GC)
C19H31BN2O2
4-4-isopropylpiperizinyl phenylboronic acid, pinacol ester, 1-isopropyl-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperazine, 4-4-isopropylpiperizinyl phenylboronic acid pinacol ester, 4-4-isopropylpiperizinyl phenylboronicacid,pinacolester, 4-4-isopropylpiperazinyl phenylboronic acid, pinacol ester, 4-4-isopropylpiperizinyl benzeneboronic acid, pinacol ester, 4-4-isopropylpiperazin-1-yl phenylboronic acid pinacol ester, 1-propan-2-yl-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperazine, 1-isopropyl-4-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperazine
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)N3CCN(CC3)C(C)C
330.28
330.28
1073354-18-3
Glass Bottle
1g
CSORKGLMGUQQOY-UHFFFAOYSA-N
1-propan-2-yl-4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazine
17750254
97%

Chemical Identifiers

1073354-18-3
330.28
4-4-isopropylpiperizinyl phenylboronic acid, pinacol ester, 1-isopropyl-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperazine, 4-4-isopropylpiperizinyl phenylboronic acid pinacol ester, 4-4-isopropylpiperizinyl phenylboronicacid,pinacolester, 4-4-isopropylpiperazinyl phenylboronic acid, pinacol ester, 4-4-isopropylpiperizinyl benzeneboronic acid, pinacol ester, 4-4-isopropylpiperazin-1-yl phenylboronic acid pinacol ester, 1-propan-2-yl-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperazine, 1-isopropyl-4-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperazine
1-propan-2-yl-4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazine
C19H31BN2O2
CSORKGLMGUQQOY-UHFFFAOYSA-N
17750254
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)N3CCN(CC3)C(C)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only