missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(4-Methoxyphenyl)butyric acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6061303

 View more versions of this product

Catalog No. AAH6061303


Explore available promotions

Description

Description

4-(4-Methoxyphenyl)butyric acid on demethylation with pyridinium hydrochloride affords 4-hydroxyphenylbutyric acid, a key starting material for the synthesis of preclinical candidate LY518674.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(4-Methoxyphenyl)butyric acid on demethylation with pyridinium hydrochloride affords 4-hydroxyphenylbutyric acid, a key starting material for the synthesis of preclinical candidate LY518674.

Solubility
Slightly soluble in methanol. Insoluble in chloroform.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

4-(4-Methoxyphenyl)butyric acid
56°C to 59°C
MFCD00004404
2416568
Slightly soluble in methanol. Insoluble in chloroform.
COC1=CC=C(C=C1)CCCC(=O)O
194.23
194.23
4521-28-2
C11H14O3
1 g
4-4-methoxyphenyl butyric acid, 4-4-methoxyphenyl butanoic acid, benzenebutanoic acid, 4-methoxy, 4-4-methoxyphenyl butyricacid, 4-4-methoxyphenyl-1-butyric acid, p-methoxy-g-phenylbutyric acid, pubchem15207, 4-methoxybenzenebutyric acid, 4-p-methoxyphenylbutyric acid
LZHMNCJMXQKSBY-UHFFFAOYSA-N
4-(4-methoxyphenyl)butanoic acid
78280
98%

Chemical Identifiers

4521-28-2
194.23
LZHMNCJMXQKSBY-UHFFFAOYSA-N
78280
COC1=CC=C(C=C1)CCCC(=O)O
C11H14O3
MFCD00004404
4-4-methoxyphenyl butyric acid, 4-4-methoxyphenyl butanoic acid, benzenebutanoic acid, 4-methoxy, 4-4-methoxyphenyl butyricacid, 4-4-methoxyphenyl-1-butyric acid, p-methoxy-g-phenylbutyric acid, pubchem15207, 4-methoxybenzenebutyric acid, 4-p-methoxyphenylbutyric acid
4-(4-methoxyphenyl)butanoic acid
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 224-849-3

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only