Learn More
4-(4-Methoxyphenyl)butyric acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6061306
Description
4-(4-Methoxyphenyl)butyric acid on demethylation with pyridinium hydrochloride affords 4-hydroxyphenylbutyric acid, a key starting material for the synthesis of preclinical candidate LY518674.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-(4-Methoxyphenyl)butyric acid on demethylation with pyridinium hydrochloride affords 4-hydroxyphenylbutyric acid, a key starting material for the synthesis of preclinical candidate LY518674.
Solubility
Slightly soluble in methanol. Insoluble in chloroform.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications
4-(4-Methoxyphenyl)butyric acid | |
56°C to 59°C | |
MFCD00004404 | |
2416568 | |
Slightly soluble in methanol. Insoluble in chloroform. | |
COC1=CC=C(C=C1)CCCC(=O)O | |
194.23 | |
194.23 |
4521-28-2 | |
C11H14O3 | |
5 g | |
4-4-methoxyphenyl butyric acid, 4-4-methoxyphenyl butanoic acid, benzenebutanoic acid, 4-methoxy, 4-4-methoxyphenyl butyricacid, 4-4-methoxyphenyl-1-butyric acid, p-methoxy-g-phenylbutyric acid, pubchem15207, 4-methoxybenzenebutyric acid, 4-p-methoxyphenylbutyric acid | |
LZHMNCJMXQKSBY-UHFFFAOYSA-N | |
4-(4-methoxyphenyl)butanoic acid | |
78280 | |
98% |
Chemical Identifiers
4521-28-2 | |
194.23 | |
LZHMNCJMXQKSBY-UHFFFAOYSA-N | |
78280 | |
COC1=CC=C(C=C1)CCCC(=O)O |
C11H14O3 | |
MFCD00004404 | |
4-4-methoxyphenyl butyric acid, 4-4-methoxyphenyl butanoic acid, benzenebutanoic acid, 4-methoxy, 4-4-methoxyphenyl butyricacid, 4-4-methoxyphenyl-1-butyric acid, p-methoxy-g-phenylbutyric acid, pubchem15207, 4-methoxybenzenebutyric acid, 4-p-methoxyphenylbutyric acid | |
4-(4-methoxyphenyl)butanoic acid |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 224-849-3
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only