missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4,4'-Methylenebis(cyclohexylamine) (mixture of isomers) 97.0+%, TCI America™
Supplier: TCI America M0699500G
Specifications
4,4′-Methylenebis(cyclohexylamine) (mixture of isomers) | |
White | |
C13H26N2 | |
500 g | |
4,4'-methylenedicyclohexanamine, 4,4'-diaminodicyclohexylmethane, 4,4'-methylenebis cyclohexylamine, cyclohexanamine, 4,4'-methylenebis, wandamin hm, bis 4-aminocyclohexyl methane, bis p-aminocyclohexyl methane, 4,4'-diaminodicyclohexyl methane, pacm 20, cyclohexylamine, 4,4'-methylenebis | |
C1CC(CCC1CC2CCC(CC2)N)N | |
210.365 | |
210.37 | |
Crystalline Lumps |
1761-71-3 | |
126°C | |
MFCD00001496 | |
2928 | |
DZIHTWJGPDVSGE-UHFFFAOYSA-N | |
4-[(4-aminocyclohexyl)methyl]cyclohexan-1-amine | |
15660 | |
≥97.0% (GC,T) |
Chemical Identifiers
1761-71-3 | |
210.365 | |
DZIHTWJGPDVSGE-UHFFFAOYSA-N | |
15660 | |
C1CC(CCC1CC2CCC(CC2)N)N |
C13H26N2 | |
MFCD00001496 | |
4,4'-methylenedicyclohexanamine, 4,4'-diaminodicyclohexylmethane, 4,4'-methylenebis cyclohexylamine, cyclohexanamine, 4,4'-methylenebis, wandamin hm, bis 4-aminocyclohexyl methane, bis p-aminocyclohexyl methane, 4,4'-diaminodicyclohexyl methane, pacm 20, cyclohexylamine, 4,4'-methylenebis | |
4-[(4-aminocyclohexyl)methyl]cyclohexan-1-amine |
Safety and Handling
EINECSNumber : (3)-2272&(4)-0101
RTECSNumber : GX1530000
TSCA : Yes