missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-(4-methylperhydro-1,4-diazepin-1-yl)benzonitrile, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC60316DA
Specifications
4-(4-Methylperhydro-1, 4-diazepin-1-yl)benzonitrile | |
97% | |
1g | |
LWRMQRYMWJWCLS-UHFFFAOYSA-N | |
4-(4-methyl-1,4-diazepan-1-yl)benzonitrile | |
18001106 | |
97% |
166438-80-8 | |
C13H17N3 | |
4-4-methyl-1,4-diazepan-1-yl benzonitrile, 4-4-methylperhydro-1,4-diazepin-1-yl benzonitrile, benzonitrile,4-hexahydro-4-methyl-1h-1,4-diazepin-1-yl, 4-4-methylhomopiperazin-1-yl benzonitrile, 4-4-methyl-1,4-diazaperhydroepinyl benzenecarbonitrile, 4-hexahydro-4-methyl-1h-1,4-diazepine-1-yl benzonitrile | |
CN1CCCN(CC1)C2=CC=C(C=C2)C#N | |
215.3 | |
215.3 |
Chemical Identifiers
166438-80-8 | |
215.3 | |
4-4-methyl-1,4-diazepan-1-yl benzonitrile, 4-4-methylperhydro-1,4-diazepin-1-yl benzonitrile, benzonitrile,4-hexahydro-4-methyl-1h-1,4-diazepin-1-yl, 4-4-methylhomopiperazin-1-yl benzonitrile, 4-4-methyl-1,4-diazaperhydroepinyl benzenecarbonitrile, 4-hexahydro-4-methyl-1h-1,4-diazepine-1-yl benzonitrile | |
4-(4-methyl-1,4-diazepan-1-yl)benzonitrile |
C13H17N3 | |
LWRMQRYMWJWCLS-UHFFFAOYSA-N | |
18001106 | |
CN1CCCN(CC1)C2=CC=C(C=C2)C#N |
Safety and Handling
GHS Signal Word: Danger