missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(4-methylperhydro-1,4-diazepin-1-yl)benzonitrile, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC60316DE

 View more versions of this product

Catalog No. CC60316DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

4-(4-Methylperhydro-1, 4-diazepin-1-yl)benzonitrile
97%
C13H17N3
4-4-methyl-1,4-diazepan-1-yl benzonitrile, 4-4-methylperhydro-1,4-diazepin-1-yl benzonitrile, benzonitrile,4-hexahydro-4-methyl-1h-1,4-diazepin-1-yl, 4-4-methylhomopiperazin-1-yl benzonitrile, 4-4-methyl-1,4-diazaperhydroepinyl benzenecarbonitrile, 4-hexahydro-4-methyl-1h-1,4-diazepine-1-yl benzonitrile
CN1CCCN(CC1)C2=CC=C(C=C2)C#N
215.3
215.3
166438-80-8
Amber glass bottle
5g
LWRMQRYMWJWCLS-UHFFFAOYSA-N
4-(4-methyl-1,4-diazepan-1-yl)benzonitrile
18001106
97%

Chemical Identifiers

166438-80-8
215.3
4-4-methyl-1,4-diazepan-1-yl benzonitrile, 4-4-methylperhydro-1,4-diazepin-1-yl benzonitrile, benzonitrile,4-hexahydro-4-methyl-1h-1,4-diazepin-1-yl, 4-4-methylhomopiperazin-1-yl benzonitrile, 4-4-methyl-1,4-diazaperhydroepinyl benzenecarbonitrile, 4-hexahydro-4-methyl-1h-1,4-diazepine-1-yl benzonitrile
4-(4-methyl-1,4-diazepan-1-yl)benzonitrile
C13H17N3
LWRMQRYMWJWCLS-UHFFFAOYSA-N
18001106
CN1CCCN(CC1)C2=CC=C(C=C2)C#N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions