missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(4-Methylpiperazino)benzylamine, ≥90%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC18713DA

 View more versions of this product

Catalog No. CC18713DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

4-(4-Methylpiperazino)benzylamine
89.5
90%
C12H19N3
1g
MZFQGKRIWIKPBT-UHFFFAOYSA-N
[4-(4-methylpiperazin-1-yl)phenyl]methanamine
2776493
90%
216144-45-5
100.0
Amber Glass Bottle
MFCD03086117
4-4-methylpiperazino benzylamine, 4-4-methylpiperazin-1-yl benzylamine, 4-4-methylpiperazin-1-yl phenyl methanamine, 1-4-4-methylpiperazin-1-yl phenyl methanamine, 4-4-methyl-piperazin-1-yl-benzylamine, 4-4-methylpiperazin-1-yl-benzylamine, benzenemethanamine, 4-4-methyl-1-piperazinyl, 4-4-methylpiperazinyl phenyl methylamine, pubchem9053
CN1CCN(CC1)C2=CC=C(C=C2)CN
205.305
205.3

Chemical Identifiers

216144-45-5
205.305
MZFQGKRIWIKPBT-UHFFFAOYSA-N
2776493
CN1CCN(CC1)C2=CC=C(C=C2)CN
C12H19N3
MFCD03086117
4-4-methylpiperazino benzylamine, 4-4-methylpiperazin-1-yl benzylamine, 4-4-methylpiperazin-1-yl phenyl methanamine, 1-4-4-methylpiperazin-1-yl phenyl methanamine, 4-4-methyl-piperazin-1-yl-benzylamine, 4-4-methylpiperazin-1-yl-benzylamine, benzenemethanamine, 4-4-methyl-1-piperazinyl, 4-4-methylpiperazinyl phenyl methylamine, pubchem9053
[4-(4-methylpiperazin-1-yl)phenyl]methanamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions