Learn More
4-(4-Morpholinyl)aniline, 98+%, Thermo Scientific Chemicals
$117.27 - $380.33
Chemical Identifiers
CAS | 2524-67-6 |
---|---|
Molecular Formula | C10H14N2O |
Molecular Weight (g/mol) | 178.235 |
MDL Number | MFCD00006169 |
InChI Key | PHNDZBFLOPIMSM-UHFFFAOYSA-N |
Synonym | 4-morpholinoaniline, 4-morpholin-4-yl-phenylamine, 4-morpholin-4-yl aniline, n-4-aminophenyl morpholine, p-morpholinoaniline, 4-morpholinobenzenamine, 4-4-morpholino aniline, benzenamine, 4-4-morpholinyl, 4-morpholinyl aniline, 4-4-aminophenyl-morpholine |
PubChem CID | 75655 |
IUPAC Name | 4-morpholin-4-ylaniline |
SMILES | C1COCCN1C2=CC=C(C=C2)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2029206
|
Thermo Scientific Chemicals
B2029206 |
5 g |
Each for $117.27
|
|
|||||
AAB2029214
|
Thermo Scientific Chemicals
B2029214 |
25 g |
Each for $380.33
|
|
|||||
Description
4-(4-Morpholinyl)aniline is used in the preparation of central nervous system (CNS) active agents, also used in the color developing process in photographs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-(4-Morpholinyl)aniline is used in the preparation of central nervous system (CNS) active agents, also used in the color developing process in photographs.
Solubility
Soluble in chloroform, ethyl acetate.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents, acid hydrides, acid chlorides, chloroformates.
Chemical Identifiers
2524-67-6 | |
178.235 | |
PHNDZBFLOPIMSM-UHFFFAOYSA-N | |
75655 | |
C1COCCN1C2=CC=C(C=C2)N |
C10H14N2O | |
MFCD00006169 | |
4-morpholinoaniline, 4-morpholin-4-yl-phenylamine, 4-morpholin-4-yl aniline, n-4-aminophenyl morpholine, p-morpholinoaniline, 4-morpholinobenzenamine, 4-4-morpholino aniline, benzenamine, 4-4-morpholinyl, 4-morpholinyl aniline, 4-4-aminophenyl-morpholine | |
4-morpholin-4-ylaniline |
Specifications
2524-67-6 | |
C10H14N2O | |
5 g | |
150957 | |
Soluble in chloroform,ethyl acetate. | |
C1COCCN1C2=CC=C(C=C2)N | |
178.235 | |
178.24 | |
4-(4-Morpholinyl)aniline |
129°C to 133°C | |
MFCD00006169 | |
UN2811 | |
4-morpholinoaniline, 4-morpholin-4-yl-phenylamine, 4-morpholin-4-yl aniline, n-4-aminophenyl morpholine, p-morpholinoaniline, 4-morpholinobenzenamine, 4-4-morpholino aniline, benzenamine, 4-4-morpholinyl, 4-morpholinyl aniline, 4-4-aminophenyl-morpholine | |
PHNDZBFLOPIMSM-UHFFFAOYSA-N | |
4-morpholin-4-ylaniline | |
75655 | |
≥98% |
Safety and Handling
GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 219-760-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only