Learn More
4',5,7-Trihydroxyisoflavone, 99+%, Thermo Scientific Chemicals
Inhibitor of tyrosine protein kinase
$52.37 - $1173.14
Chemical Identifiers
CAS | 446-72-0 |
---|---|
Molecular Formula | C15H10O5 |
Molecular Weight (g/mol) | 270.24 |
MDL Number | MFCD00016952 |
InChI Key | TZBJGXHYKVUXJN-UHFFFAOYSA-N |
Synonym | genistein, prunetol, genisteol, 4',5,7-trihydroxyisoflavone, genisterin, sophoricol, 5,7,4'-trihydroxyisoflavone, bonistein, 5,7-dihydroxy-3-4-hydroxyphenyl-4h-chromen-4-one, genestein |
PubChem CID | 5280961 |
ChEBI | CHEBI:28088 |
IUPAC Name | 5,7-dihydroxy-3-(4-hydroxyphenyl)chromen-4-one |
SMILES | OC1=CC=C(C=C1)C1=COC2=CC(O)=CC(O)=C2C1=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ63241MC
|
Thermo Scientific Chemicals
J63241MC |
100 mg |
Each for $52.37
|
|
|||||
AAJ6324103
|
Thermo Scientific Chemicals
J6324103 |
1 g |
Each for $288.00
|
|
|||||
AAJ6324109
|
Thermo Scientific Chemicals
J6324109 |
10 g |
Each for $1,173.14
|
|
|||||
Description
4',5,7-Trihydroxyisoflavone is used to inhibitor of tyrosine protein kinase. Genistein inhibits EGF-stimulated phosphorylation in cultured cells. Also used to explore signal transduction pathways.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4′,5,7-Trihydroxyisoflavone is used to inhibitor of tyrosine protein kinase. Genistein inhibits EGF-stimulated phosphorylation in cultured cells. Also used to explore signal transduction pathways.
Solubility
Soluble in chloroform, methanol. Insoluble in cold water.
Notes
Protect from heat. Store away from oxidizing agents.
Chemical Identifiers
446-72-0 | |
270.24 | |
TZBJGXHYKVUXJN-UHFFFAOYSA-N | |
5280961 | |
5,7-dihydroxy-3-(4-hydroxyphenyl)chromen-4-one |
C15H10O5 | |
MFCD00016952 | |
genistein, prunetol, genisteol, 4',5,7-trihydroxyisoflavone, genisterin, sophoricol, 5,7,4'-trihydroxyisoflavone, bonistein, 5,7-dihydroxy-3-4-hydroxyphenyl-4h-chromen-4-one, genestein | |
CHEBI:28088 | |
OC1=CC=C(C=C1)C1=COC2=CC(O)=CC(O)=C2C1=O |
Specifications
446-72-0 | |
Brown to White | |
MFCD00016952 | |
263823 | |
genistein, prunetol, genisteol, 4',5,7-trihydroxyisoflavone, genisterin, sophoricol, 5,7,4'-trihydroxyisoflavone, bonistein, 5,7-dihydroxy-3-4-hydroxyphenyl-4h-chromen-4-one, genestein | |
TZBJGXHYKVUXJN-UHFFFAOYSA-N | |
5,7-dihydroxy-3-(4-hydroxyphenyl)chromen-4-one | |
5280961 | |
270.24 | |
Powder |
∼300°C | |
C15H10O5 | |
100 mg | |
14,4391 | |
Soluble in chloroform,methanol. Insoluble in cold water. | |
OC1=CC=C(C=C1)C1=COC2=CC(O)=CC(O)=C2C1=O | |
270.24 | |
CHEBI:28088 | |
≥99% | |
4',5,7-Trihydroxyisoflavone |
Safety and Handling
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 207-174-9
RTECSNumber : NR2392000
TSCA : Yes
Recommended Storage : Store at -20°C
RUO – Research Use Only