missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4,5-Diamino-6-hydroxypyrimidine 96.0+%, TCI America™
$70.67 - $257.46
Chemical Identifiers
CAS | 1672-50-0 |
---|---|
Molecular Formula | C4H6N4O |
Molecular Weight (g/mol) | 126.12 |
MDL Number | MFCD00006114 |
InChI Key | PWRHKLKFADDKHS-UHFFFAOYSA-N |
Synonym | 5,6-Diamino-4(1H)-pyrimidinone, 5,6-Diamino-4-pyrimidinol, 5,6-Diamino-4-hydroxypyrimidine |
PubChem CID | 74287 |
IUPAC Name | 5,6-diamino-1,4-dihydropyrimidin-4-one |
SMILES | NC1=C(N)C(=O)N=CN1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
D4488200MG
|
TCI America
D4488200MG |
200 mg |
Each for $70.67
|
|
|||||
D44881G
|
TCI America
D44881G |
1 g |
Each for $257.46
|
|
|||||
Chemical Identifiers
1672-50-0 | |
126.12 | |
PWRHKLKFADDKHS-UHFFFAOYSA-N | |
74287 | |
NC1=C(N)C(=O)N=CN1 |
C4H6N4O | |
MFCD00006114 | |
5,6-Diamino-4(1H)-pyrimidinone, 5,6-Diamino-4-pyrimidinol, 5,6-Diamino-4-hydroxypyrimidine | |
5,6-diamino-1,4-dihydropyrimidin-4-one |
Specifications
1672-50-0 | |
C4H6N4O | |
200 mg | |
PWRHKLKFADDKHS-UHFFFAOYSA-N | |
5,6-diamino-1,4-dihydropyrimidin-4-one | |
74287 | |
≥96.0% (HPLC,T) | |
4,5-Diamino-6-hydroxypyrimidine |
244°C | |
MFCD00006114 | |
5,6-Diamino-4(1H)-pyrimidinone, 5,6-Diamino-4-pyrimidinol, 5,6-Diamino-4-hydroxypyrimidine | |
NC1=C(N)C(=O)N=CN1 | |
126.12 | |
126.12 | |
Crystalline Powder |
Safety and Handling
TSCA : No