missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,5-Dibromo-2-phenyl-2,3-dihydropyridazin-3-one, 97%, Thermo Scientific™

Chemical Identifiers

CAS 14305-08-9
Molecular Formula C10H6Br2N2O
Molecular Weight (g/mol) 329.979
MDL Number MFCD00052907
InChI Key NQJXEMRMTOSSBQ-UHFFFAOYSA-N
Synonym 4,5-dibromo-2-phenylpyridazin-3 2h-one, 4,5-dibromo-2-phenyl-2,3-dihydropyridazin-3-one, 4,5-dibromo-2-phenyl-3 2h-pyridazinone, 3 2h-pyridazinone, 4,5-dibromo-2-phenyl, 4,5-dibromo-2-phenyl-3-pyridazinone, 4,5-dibromo-2-phenyl-pyridazin-3-one, 2-phenyl-4,5-dibromo-3 2h-pyridazinone, 3 2h-pyridazinone,4,5-dibromo-2-phenyl, 4,5-bis bromanyl-2-phenyl-pyridazin-3-one, 4,5-dibromo-2-phenyl-2-hydropyridazin-3-one
PubChem CID 203396
IUPAC Name 4,5-dibromo-2-phenylpyridazin-3-one
SMILES C1=CC=C(C=C1)N2C(=O)C(=C(C=N2)Br)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BTB07379DA
SDS View Documents
Thermo Scientific Chemicals
BTB07379DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTB07379EA
SDS View Documents
Thermo Scientific Chemicals
BTB07379EA
10g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

14305-08-9
329.979
NQJXEMRMTOSSBQ-UHFFFAOYSA-N
203396
C1=CC=C(C=C1)N2C(=O)C(=C(C=N2)Br)Br
C10H6Br2N2O
MFCD00052907
4,5-dibromo-2-phenylpyridazin-3 2h-one, 4,5-dibromo-2-phenyl-2,3-dihydropyridazin-3-one, 4,5-dibromo-2-phenyl-3 2h-pyridazinone, 3 2h-pyridazinone, 4,5-dibromo-2-phenyl, 4,5-dibromo-2-phenyl-3-pyridazinone, 4,5-dibromo-2-phenyl-pyridazin-3-one, 2-phenyl-4,5-dibromo-3 2h-pyridazinone, 3 2h-pyridazinone,4,5-dibromo-2-phenyl, 4,5-bis bromanyl-2-phenyl-pyridazin-3-one, 4,5-dibromo-2-phenyl-2-hydropyridazin-3-one
4,5-dibromo-2-phenylpyridazin-3-one

Specifications

14305-08-9
100.0
Amber Glass Bottle
MFCD00052907
4,5-dibromo-2-phenylpyridazin-3 2h-one, 4,5-dibromo-2-phenyl-2,3-dihydropyridazin-3-one, 4,5-dibromo-2-phenyl-3 2h-pyridazinone, 3 2h-pyridazinone, 4,5-dibromo-2-phenyl, 4,5-dibromo-2-phenyl-3-pyridazinone, 4,5-dibromo-2-phenyl-pyridazin-3-one, 2-phenyl-4,5-dibromo-3 2h-pyridazinone, 3 2h-pyridazinone,4,5-dibromo-2-phenyl, 4,5-bis bromanyl-2-phenyl-pyridazin-3-one, 4,5-dibromo-2-phenyl-2-hydropyridazin-3-one
C1=CC=C(C=C1)N2C(=O)C(=C(C=N2)Br)Br
329.979
329.98
4, 5-Dibromo-2-phenyl-2, 3-dihydropyridazin-3-one
96.5
97%
C10H6Br2N2O
1g
NQJXEMRMTOSSBQ-UHFFFAOYSA-N
4,5-dibromo-2-phenylpyridazin-3-one
203396
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'