Learn More
4,5-Dibromothiophene-2-sulfonyl chloride, 97%
CAS: 81606-31-7 | C4HBr2ClO2S2 | 340.43 g/mol
$298.34 - $1,230.42
Chemical Identifiers
| CAS | 81606-31-7 |
|---|---|
| Molecular Formula | C4HBr2ClO2S2 |
| Molecular Weight (g/mol) | 340.43 |
| MDL Number | MFCD00041439 |
| InChI Key | WJYGHWXWQSCONR-UHFFFAOYSA-N |
| Synonym | 4,5-dibromothiophene-2-sulphonyl chloride, 2,3-dibromothiophene-5-sulfonyl chloride, 4,5-dibromo-2-thiophenesulfonyl chloride, 2-thiophenesulfonyl chloride, 4,5-dibromo, 4,5-dibromothiophene-2-sulfonic acid chloride, acmc-20ams9, buttpark 9350-57, 4,5-dibromo 2-thienyl chlorosulfone, 2,3-dibromothiophene-5-sulfonylchloride, 4,5-dibromothiophene-2-sulfonylchloride |
| PubChem CID | 522755 |
| IUPAC Name | 4,5-dibromothiophene-2-sulfonyl chloride |
| SMILES | ClS(=O)(=O)C1=CC(Br)=C(Br)S1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1338406
|
Thermo Scientific Chemicals
A1338406 |
5 g |
N/A
|
|
|||||
|
AAA1338414
|
Thermo Scientific Chemicals
A1338414 |
25 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 81606-31-7 | |
| 340.43 | |
| WJYGHWXWQSCONR-UHFFFAOYSA-N | |
| 522755 | |
| ClS(=O)(=O)C1=CC(Br)=C(Br)S1 |
| C4HBr2ClO2S2 | |
| MFCD00041439 | |
| 4,5-dibromothiophene-2-sulphonyl chloride, 2,3-dibromothiophene-5-sulfonyl chloride, 4,5-dibromo-2-thiophenesulfonyl chloride, 2-thiophenesulfonyl chloride, 4,5-dibromo, 4,5-dibromothiophene-2-sulfonic acid chloride, acmc-20ams9, buttpark 9350-57, 4,5-dibromo 2-thienyl chlorosulfone, 2,3-dibromothiophene-5-sulfonylchloride, 4,5-dibromothiophene-2-sulfonylchloride | |
| 4,5-dibromothiophene-2-sulfonyl chloride |
Specifications
| 81606-31-7 | |
| C4HBr2ClO2S2 | |
| 5 g | |
| 162960 | |
| 4,5-dibromothiophene-2-sulphonyl chloride, 2,3-dibromothiophene-5-sulfonyl chloride, 4,5-dibromo-2-thiophenesulfonyl chloride, 2-thiophenesulfonyl chloride, 4,5-dibromo, 4,5-dibromothiophene-2-sulfonic acid chloride, acmc-20ams9, buttpark 9350-57, 4,5-dibromo 2-thienyl chlorosulfone, 2,3-dibromothiophene-5-sulfonylchloride, 4,5-dibromothiophene-2-sulfonylchloride | |
| ClS(=O)(=O)C1=CC(Br)=C(Br)S1 | |
| 340.43 | |
| 340.45 | |
| 4,5-Dibromothiophene-2-sulfonyl chloride |
| 75°C to 79°C | |
| MFCD00041439 | |
| UN3261 | |
| Moisture sensitive | |
| WJYGHWXWQSCONR-UHFFFAOYSA-N | |
| 4,5-dibromothiophene-2-sulfonyl chloride | |
| 522755 | |
| 97% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Hazard Class: 8; Packaging Group: III
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only