missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
4,5-Dichloro-2-(4-methylphenyl)-2,3-dihydropyridazin-3-one, 97%, Thermo Scientific™
Chemical Identifiers
CAS
33098-21-4
Molecular Formula
C11H8Cl2N2O
Molecular Weight (g/mol)
255.098
MDL Number
MFCD00135290
InChI Key
OWPCMQKRJFNLHF-UHFFFAOYSA-N
Synonym
4,5-dichloro-2-4-methylphenyl-2,3-dihydropyridazin-3-one, 4,5-dichloro-2-4-methylphenyl pyridazin-3-one, 3 2h-pyridazinone,4,5-dichloro-2-4-methylphenyl, 4,5-dichloro-2-p-tolyl-2h-pyridazin-3-one, 4,5-dichloro-2-4-methylphenyl-2-hydropyridazin-3-one, 4,5-dichloro-2-p-tolyl pyridazin-3 2h-one, 4,5-dichloro-2-4-methylphenyl-3 2h-pyridazinone, 4,5-dichloro-2-4-methylphenyl pyridazin-3 2h-one, 4,5-dichloro-2-4-methylphenyl-2,3-dihydropyridazin-3one
PubChem CID
2774757
IUPAC Name
4,5-dichloro-2-(4-methylphenyl)pyridazin-3-one
SMILES
CC1=CC=C(C=C1)N2C(=O)C(=C(C=N2)Cl)Cl
View More Specs
Products
2
Description
Specifications
Chemical Identifiers
CAS
33098-21-4
Molecular Weight (g/mol)
255.098
InChI Key
OWPCMQKRJFNLHF-UHFFFAOYSA-N
PubChem CID
2774757
SMILES
CC1=CC=C(C=C1)N2C(=O)C(=C(C=N2)Cl)Cl
Molecular Formula
C11H8Cl2N2O
MDL Number
MFCD00135290
Synonym
4,5-dichloro-2-4-methylphenyl-2,3-dihydropyridazin-3-one, 4,5-dichloro-2-4-methylphenyl pyridazin-3-one, 3 2h-pyridazinone,4,5-dichloro-2-4-methylphenyl, 4,5-dichloro-2-p-tolyl-2h-pyridazin-3-one, 4,5-dichloro-2-4-methylphenyl-2-hydropyridazin-3-one, 4,5-dichloro-2-p-tolyl pyridazin-3 2h-one, 4,5-dichloro-2-4-methylphenyl-3 2h-pyridazinone, 4,5-dichloro-2-4-methylphenyl pyridazin-3 2h-one, 4,5-dichloro-2-4-methylphenyl-2,3-dihydropyridazin-3one
IUPAC Name
4,5-dichloro-2-(4-methylphenyl)pyridazin-3-one
Specifications
CAS
33098-21-4
CAS Max %
100.0
Molecular Formula
C11H8Cl2N2O
Quantity
1g
InChI Key
OWPCMQKRJFNLHF-UHFFFAOYSA-N
IUPAC Name
4,5-dichloro-2-(4-methylphenyl)pyridazin-3-one
PubChem CID
2774757
Percent Purity
97%
CAS Min %
96.5
Assay Percent Range
97%
MDL Number
MFCD00135290
Synonym
4,5-dichloro-2-4-methylphenyl-2,3-dihydropyridazin-3-one, 4,5-dichloro-2-4-methylphenyl pyridazin-3-one, 3 2h-pyridazinone,4,5-dichloro-2-4-methylphenyl, 4,5-dichloro-2-p-tolyl-2h-pyridazin-3-one, 4,5-dichloro-2-4-methylphenyl-2-hydropyridazin-3-one, 4,5-dichloro-2-p-tolyl pyridazin-3 2h-one, 4,5-dichloro-2-4-methylphenyl-3 2h-pyridazinone, 4,5-dichloro-2-4-methylphenyl pyridazin-3 2h-one, 4,5-dichloro-2-4-methylphenyl-2,3-dihydropyridazin-3one
SMILES
CC1=CC=C(C=C1)N2C(=O)C(=C(C=N2)Cl)Cl
Molecular Weight (g/mol)
255.098
Formula Weight
255.1
Chemical Name or Material
4, 5-Dichloro-2-(4-methylphenyl)-2, 3-dihydropyridazin-3-one
Videos
Safety and Handling
Safety and Handling
GHS H Statement: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
GHS P Statement: Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
SDS
missing translation for 'documents'