Learn More
4,5-Dichloro-2-methyl-3(2H)-pyridazinone, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 399710010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4, 5-Dichloro-2-methyl-3(2H)-pyridazinone | |
87.0°C to 91.0°C | |
Glass bottle | |
MFCD00051686 | |
4,5-dichloro-2-methylpyridazin-3 2h-one, 4,5-dichloro-2-methyl-3 2h-pyridazinone, 3 2h-pyridazinone, 4,5-dichloro-2-methyl, 4,5-dichloro-2-methyl-2h-pyridazin-3-one, 4,5-dichloro-2-methyl-2,3-dihydropyridazin-3-one, 3 2h-pyridazinone,4,5-dichloro-2-methyl, 4,5-dichloro-2-methyl-2-hydropyridazin-3-one, pubchem16243, acmc-209wiz, 5-24-02-00023 beilstein handbook reference | |
CN1N=CC(Cl)=C(Cl)C1=O | |
179.00 | |
179.01 |
933-76-6 | |
96% min. (GC) | |
C5H4Cl2N2O | |
1 g | |
ACKBTCUMGAHRIE-UHFFFAOYSA-N | |
4,5-dichloro-2-methylpyridazin-3-one | |
120462 | |
97% |
Chemical Identifiers
933-76-6 | |
179.00 | |
ACKBTCUMGAHRIE-UHFFFAOYSA-N | |
120462 | |
CN1N=CC(Cl)=C(Cl)C1=O |
C5H4Cl2N2O | |
MFCD00051686 | |
4,5-dichloro-2-methylpyridazin-3 2h-one, 4,5-dichloro-2-methyl-3 2h-pyridazinone, 3 2h-pyridazinone, 4,5-dichloro-2-methyl, 4,5-dichloro-2-methyl-2h-pyridazin-3-one, 4,5-dichloro-2-methyl-2,3-dihydropyridazin-3-one, 3 2h-pyridazinone,4,5-dichloro-2-methyl, 4,5-dichloro-2-methyl-2-hydropyridazin-3-one, pubchem16243, acmc-209wiz, 5-24-02-00023 beilstein handbook reference | |
4,5-dichloro-2-methylpyridazin-3-one |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persist
GHS Signal Word: Danger
RUO – Research Use Only