missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,5-Dicyanoimidazole, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 368530050

 View more versions of this product

Catalog No. AC368530050

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4, 5-Dicyanoimidazole
1122-28-7
100.0
White
98.5% min. (HPLC)
C5H2N4
5 g
XGDRLCRGKUCBQL-UHFFFAOYSA-N
1H-imidazole-4,5-dicarbonitrile
70729
99%
99%
98.5
174.0°C to 177.0°C
Authentic
Glass bottle
MFCD00005194
4,5-dicyanoimidazole, imidazole-4,5-dicarbonitrile, 4,5-imidazoledicarbonitrile, activator, pubchem7602, 4, 5-dicyanoimidazole, moa 7, imidazole-4,5-dinitrile, acmc-1brr7, 4,5-dicyano-1h-imidazole
N#CC1=C(N=CN1)C#N
118.10
118.1
Crystalline Powder

Chemical Identifiers

1122-28-7
118.10
XGDRLCRGKUCBQL-UHFFFAOYSA-N
1H-imidazole-4,5-dicarbonitrile
C5H2N4
MFCD00005194
70729
N#CC1=C(N=CN1)C#N
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Immediately call a POISON CENTER or

GHS Signal Word: Danger

EINECSNumber : 214-344-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only