missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4,5-Difluoro-2-nitroaniline 98.0+%, TCI America™
Supplier: TCI America D364425G
Specifications
4,5-Difluoro-2-nitroaniline | |
108°C | |
MFCD00010876 | |
3,4-difluoro-6-nitroaniline, benzenamine, 4,5-difluoro-2-nitro, 4,5-difluoro-2-nitro-aniline, 4,5-difluoro-2-nitrophenylamine, 2-nitro-4,5-difluoroaniline, 2-amino-4,5-difluoronitrobenzene, 4,5-difluoro-2-nitro-phenylamine, 4,5-difluoro-2-nitrobenzenamine, pubchem2121, acmc-209pdb | |
NC1=CC(F)=C(F)C=C1[N+]([O-])=O | |
174.11 | |
174.11 | |
Crystalline Powder |
78056-39-0 | |
C6H4F2N2O2 | |
25 g | |
WDMCABATCGQAKK-UHFFFAOYSA-N | |
4,5-difluoro-2-nitroaniline | |
157151 | |
≥98.0% (GC) |
Chemical Identifiers
78056-39-0 | |
174.11 | |
WDMCABATCGQAKK-UHFFFAOYSA-N | |
157151 | |
NC1=CC(F)=C(F)C=C1[N+]([O-])=O |
C6H4F2N2O2 | |
MFCD00010876 | |
3,4-difluoro-6-nitroaniline, benzenamine, 4,5-difluoro-2-nitro, 4,5-difluoro-2-nitro-aniline, 4,5-difluoro-2-nitrophenylamine, 2-nitro-4,5-difluoroaniline, 2-amino-4,5-difluoronitrobenzene, 4,5-difluoro-2-nitro-phenylamine, 4,5-difluoro-2-nitrobenzenamine, pubchem2121, acmc-209pdb | |
4,5-difluoro-2-nitroaniline |
Safety and Handling
TSCA : No